Skip to Content
Merck
All Photos(1)

Key Documents

P6056

Sigma-Aldrich

Endoproteinase Arg-C from mouse submaxillary gland

suitable for protein sequencing, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

form

lyophilized powder

Quality Level

packaging

vial of 5 μg

suitability

suitable for protein sequencing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Endoproteinase Arg-C from mouse submaxillary gland has been used to study the syncytium formation in MERS-CoV (middle East respiratory syndrome coronavirus)-infected Vero cells in the presence of exogenous proteases. It has been used for the digestion of Rpl23ab (ribosomal protein L23ab)-containing fraction for LC-MS (liquid chromatography–mass spectrometry)/MS analysis.

Biochem/physiol Actions

Endoproteinase Arg-C is a serine endoprotease from mouse submaxillary gland which hydrolyzes peptide bonds at the carboxyl side of arginyl residues. The enzyme has been shown to cleave Lys-Lys and Lys-Arg bonds, and all Arg-X bonds may not be hydrolyzed.

Unit Definition

One unit will hydrolyze 1.0 μmole of Nα-p-tosyl-L-arginine methyl ester per min at pH 8.0 at 25 °C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Casas-Finet et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(3), 1050-1054 (1992-02-01)
To identify the functional residues of the N-terminal B region of bacteriophage T4 gene 32 protein involved in its cooperative binding to single-stranded nucleic acids, a process dependent on homotypic protein-protein interaction, we have studied the interaction of the protein
Proteomics in Functional Genomics: Protein Structure Analysis (2000)
Middle East respiratory syndrome coronavirus infection mediated by the transmembrane serine protease TMPRSS2.
Shirato K, et al.
Journal of Virology, 87, 12552-12552 (2013)
Analysis of endoproteinase Arg C action on adrenocorticotrophic hormone by capillary electrophoresis and reversed-phase high-performance liquid chromatography.
Krueger RJ, et al.
Journal of Chromatography A, 543, 451-461 (1991)
Leesa J Deterding et al.
Journal of proteome research, 7(6), 2368-2379 (2008-04-18)
Global changes in the phosphorylation state of human H1 isoforms isolated from UL3 cells have been investigated using mass spectrometry. Relative changes in H1 phosphorylation between untreated cells and cells treated with dexamethasone or various CDK inhibitors were determined. The

Protocols

An optimized LC-MS/MS based workflow for low artifact tryptic digestion and peptide mapping of monoclonal antibody, adalimumab (Humira) using filter assisted sample preparation (FASP).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service