Skip to Content
Merck
All Photos(3)

Key Documents

P5884

Sigma-Aldrich

Brij® 58

average Mn ~1124

Synonym(s):

Polyethylene glycol hexadecyl ether, Polyoxyethylene (20) cetyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)20C16H33
CAS Number:
Beilstein:
1899885
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

form

solid

mol wt

micellar avg mol wt 79,000
average Mn ~1124

aggregation number

70

technique(s)

protein quantification: suitable

CMC

0.08 mM (20-25°C)

mp

~38 °C

transition temp

cloud point >100 °C

HLB

16

SMILES string

CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C56H114O21/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-58-19-21-60-23-25-62-27-29-64-31-33-66-35-37-68-39-41-70-43-45-72-47-49-74-51-53-76-55-56-77-54-52-75-50-48-73-46-44-71-42-40-69-38-36-67-34-32-65-30-28-63-26-24-61-22-20-59-18-16-57/h57H,2-56H2,1H3

InChI key

NLMKTBGFQGKQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brij 58 is a nonionic surfactant or detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts.

Application

Polyethylene glycol hexadecyl ether, or Brij 58, has been used in a study to assess properties of gemini-conventional surfactant mixtures and their effects on solubilization of polycyclic aromatic hydrocarbons. It has also been used in a study to investigate RNase-sensitive constraint of bacterial nucleoid structure.

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kellermayer et al.
Journal of cellular physiology, 159(2), 197-204 (1994-05-01)
The plasma membrane of erythrocytes, as of other cells, is thought to act as the barrier responsible for maintaining intracellular gradients of most ions and small molecular species between the cell and its environment. Controlled application of the nonionic detergent
Macromolecular crowding can account for RNase-sensitive constraint of bacterial nucleoid structure
Foley, P., et al
Biochemical and Biophysical Research Communications, 395, 6-6 (2010)
Study of surface and solution properties of gemini-conventional surfactant mixtures and their effects on solubilization of polycyclic aromatic hydrocarbons
Panda
Journal of Molecular Liquids, 163, 6-6 (2011)
Photodestruction of BDE-99 in micellar solutions of nonionic surfactants of Brij 35 and Brij 58
Li, X., et al.
Chemosphere, 78, 8-8 (2010)
Preparation of extracts from prokaryotes.
M Cull et al.
Methods in enzymology, 182, 147-153 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service