Skip to Content
Merck
All Photos(1)

Key Documents

P5648

Sigma-Aldrich

β-Propiolactone

Grade II, ≥90%

Synonym(s):

Oxetan-2-one, 3-Hydroxypropionic acid lactone, Hydracrylic acid β-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O2
CAS Number:
Molecular Weight:
72.06
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.77

type

Grade II

Quality Level

Assay

≥90%

refractive index

n20/D 1.412 (lit.)

bp

162 °C (lit.)

mp

−33 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

O=C1CCO1

InChI

1S/C3H4O2/c4-3-1-2-5-3/h1-2H2

InChI key

VEZXCJBBBCKRPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Propiolactone is an alkylating agent which is mainly used in veterinary vaccines. It will not interfere with the antigenicity of vaccine.

Application

β-Propiolactone has been used for Influenza virus deactivation.

Caution

Insoluble polymers may form during storage.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tiago Abreu-Mota et al.
Nature communications, 9(1), 4223-4223 (2018-10-13)
Lassa fever (LF), caused by Lassa virus (LASV), is a viral hemorrhagic fever for which no approved vaccine or potent antiviral treatment is available. LF is a WHO priority disease and, together with rabies, a major health burden in West
Plasmonics-Based Detection of Virus Using Sialic Acid Functionalized Gold Nanoparticles
Biosensors and Biodetection, 109-116 (2017)
Drishya Kurup et al.
PLoS pathogens, 17(3), e1009383-e1009383 (2021-03-26)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is an emergent coronavirus that has caused a worldwide pandemic. Although human disease is often asymptomatic, some develop severe illnesses such as pneumonia, respiratory failure, and death. There is an urgent need for
Vesla I Kullaya et al.
Viral immunology, 30(8), 576-581 (2017-08-08)
Detection of respiratory syncytial virus (RSV) in blood, including mononuclear leukocytes and organs other than the lung, suggests that RSV disseminates outside the respiratory tract. In this study, the role of platelets in host defense against RSV was explored using
Veterinary Immunology-E book, 262-262 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service