Skip to Content
Merck
All Photos(1)

Key Documents

P5188

Sigma-Aldrich

TWEEN® 80

for molecular biology, syrup

Synonym(s):

POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

grade

for molecular biology

Quality Level

description

non-ionic

form

syrup

mol wt

average mol wt 1310

composition

Oleic acid, ≥58.0% (balance primarily linoleic, palmitic, and stearic acids)

CMC

0.012 mM

transition temp

cloud point 65 °C

density

1.06 g/mL at 25 °C

HLB

15

suitability

molecular biology tested

foreign activity

DNAse (Exonuclease Detection), NICKase (Endonuclease Detection), RNAse, none detected

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TWEEN 80 is a polyethylene sorbitol ester and has a molecular weight of 1.31 kDa. It is useful as a stabilizer and emulsifier commercially. It is known to be used in ice-cream for its drying property.

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fats in Food Technology (2014)
Wei-Ju Chung et al.
Journal of biological physics, 43(4), 511-524 (2017-09-10)
DNA samples are commonly frozen for storage. However, freezing can compromise the integrity of DNA molecules. Considering the wide applications of DNA molecules in nanotechnology, changes to DNA integrity at the molecular level may cause undesirable outcomes. However, the effects
Ice Cream, 86-86 (2003)
Yun Xia et al.
Cell, 175(4), 1059-1073 (2018-10-03)
Motivated by the clinical observation that interruption of the mevalonate pathway stimulates immune responses, we hypothesized that this pathway may function as a druggable target for vaccine adjuvant discovery. We found that lipophilic statin drugs and rationally designed bisphosphonates that
Cristina Marcos-Arias et al.
BMC complementary and alternative medicine, 11, 119-119 (2011-11-29)
Candida-associated denture stomatitis is a frequent infectious disease. Treatment of this oral condition is difficult because failures and recurrences are common. The aim of this study was to test the in vitro antifungal activity of pure constituents of essentials oils.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service