Skip to Content
Merck
All Photos(1)

Key Documents

P3326

Sigma-Aldrich

Platelet-Derived Growth Factor-AB human

≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

PDGF-AB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Platelet-Derived Growth Factor-AB human, PDGF-AB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE and N-terminal analysis)

form

lyophilized powder

potency

10-100 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

27-31 kDa

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Platelet-Derived Growth Factor (PDGF) is a cationic polypeptide and a constituent of α-granules. PDGF is composed of chain A and B linked together by a disulphide bridge. PDGF heterodimers occurs as PDGF-AB and homodimers exists as FDGF-AA, PDGF-BB, PDGF-CC and PDGF-DD). The PDGFB gene is localized to pericytes in the brain and is mapped to human chromosome 7p22.3.

Application

Platelet-Derived Growth Factor-AB human has been used in luciferase reporter assay.

Biochem/physiol Actions

Cellular actions of platelet-derived growth factor (PDGF) include chemoattraction and activation of neutrophils, monocytes, and fibroblasts. PDGF appears to play a crucial role in the cellular response to tissue injury and is critically involved in the pathological progression of atherosclerosis.
PDGF stimulates fibroblast proliferation and extracellular matrix production in order to facilitate tissue repair. PDGF-AB isomer is found to be associated with angiogenesis and proliferation. Loss of PDGFB gene function induces microvascular calcification due to age-dependent phenotypical changes.

Other Notes

Heterodimer of the A and B chains of PDGF disulfide linked.

Physical form

Lyophilized from a 0.2 μm filtered solution in 40% acetonitrile and 0.1% trifluoroacetic acid.

Analysis Note

The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in quiescent NR6R-3T3 fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Injury induces in vivo expression of platelet-derived growth factor (PDGF) and PDGF receptor mRNAs in skin epithelial cells and PDGF mRNA in connective tissue fibroblasts.
Antoniades HN, et al.
Proceedings of the National Academy of Sciences of the USA, 88.2 , 565-569 (1991)
Increased levels of platelet-derived growth factor in vitreous fluid of patients with proliferative diabetic retinopathy.
Freyberger H, et al.
Experimental and Clinical Endocrinology & Diabetes, 108.02, 106-109 (2000)
Clinical Findings Associated with a De Novo Partial Trisomy 10p11. 22p15. 3 and Monosomy 7p22. 3 Detected by Chromosomal Microarray Analysis.
Kohannim O, et al.
Case Reports in Genetics, 2011 (2011)
Makoto Hiromura et al.
The Journal of biological chemistry, 281(38), 27753-27764 (2006-07-13)
The serine/threonine kinase, Akt (protein kinase B) plays a central role in the regulation of intracellular cell survival. Recently, we demonstrated that the proto-oncogene TCL1, overexpressed in human T-cell prolymphocytic leukemia, is an Akt kinase co-activator. Tightly restricted TCL1 gene
Role of platelets in the development of atherosclerosis.
Huo Y, et al.
Trends in Cardiovascular Medicine, 14.1, 18-22 (2004)

Articles

Platelet-Derived Growth Factor is the principal mitogen found in mammalian serum and is released from platelets during clot formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service