Merck
  • Identification of nerve growth factor-responsive element of the TCL1 promoter as a novel negative regulatory element.
Identification of nerve growth factor-responsive element of the TCL1 promoter as a novel negative regulatory element.
The Journal of biological chemistry (2006-07-13)
Makoto Hiromura, Futoshi Suizu, Masumi Narita, Keiichi Kinowaki, Masayuki Noguchi
ABSTRACT
The serine/threonine kinase, Akt (protein kinase B) plays a central role in the regulation of intracellular cell survival. Recently, we demonstrated that the proto-oncogene TCL1, overexpressed in human T-cell prolymphocytic leukemia, is an Akt kinase co-activator. Tightly restricted TCL1 gene expression in early developmental cells suggested that the TCL1 gene is regulated at a transcriptional level. To characterize how TCL1 gene expression is regulated, we cloned the 5'-promoter of the TCL1 gene located at human chromosome 14q32. The 5'-TCL1 promoter region contains a TATA box with cis-regulatory elements for Nur77/NGFI-B (nerve growth factor-responsive element (NBRE), CCAAGGTCA), NFkappaB, and fork head transcription factor. Nur77/NGFI-B, an orphan receptor superfamily transcription factor implicated in T-cell apoptosis, is a substrate for Akt. We hypothesized that TCL1 transactivity is regulated through Akt-induced phosphorylation of Nur77/NGFI-B in vivo. In an electrophoretic mobility shift assay with chromosomal immunoprecipitation assays, wild-type Nur77, but not S350A mutant Nur77, could specifically bind to TCL1-NBRE. A luciferase assay demonstrated that TCL1-NBRE is required for inhibition of TCL1 transactivity upon nerve growth factor/platelet-derived growth factor stimulation, which activates Akt and phosphorylates Nur77. Using a chromosomal immunoprecipitation assay with reverse transcription-PCR, nerve growth factor stimulation inhibited binding of endogenous Nur77 to TCL1-NBRE, in turn, suppressing TCL1 gene expression. The results together establish that TCL1-NBRE is a novel negative regulatory element of Nur77 (NGFI-B). To the best of our knowledge, TCL1-NBRE is the first direct target of Nur77 involving the regulation of intracellular cell death survival. This Akt-induced inhibitory mechanism of TCL1 should play an important role in immunological and/or neuronal development in vivo.
Sigma-Aldrich
Platelet-Derived Growth Factor-AB human, PDGF-AB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Platelet-Derived Growth Factor-BB from rat, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture