Skip to Content
Merck
All Photos(1)

Key Documents

P1236

Sigma-Aldrich

Protease from Bacillus amyloliquefaciens

liquid, ≥0.8 U/g

Synonym(s):

Neutrase® 0.8L

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Bacillus sp. (Bacillus amyloliquefaciens)

Quality Level

form

liquid

specific activity

≥0.8 U/g

storage temp.

2-8°C

General description

Bacterial protease produced by submerged fermentation of a selected strain of Bacillus amyloliquefaciens

Application

Protease from Bacillus amyloliquefaciens has been used for the unhairing of hides and skins. [1] It has also been used in a study to investigate peptide bond formation using the carbamoylmethyl ester as the acyl donor. [2]

Biochem/physiol Actions

Secretion of protease by Bacillus amyloliquefaciens can be inhibited by treatment with the fatty acid synthetase inhibitor cerulenin. [3]

Legal Information

A product of Novozyme Corp.
Neutrase is a registered trademark of Novozymes Corp.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keqin Ou et al.
Journal of agricultural and food chemistry, 58(8), 4894-4900 (2010-03-17)
Effects of enzymatic hydrolysates of whey protein concentrates (WPC) on iron absorption were studied using in vitro digestion combined with Caco-2 cell models for improved iron absorption. Neutrase- and papain-treated WPC could improve iron absorption; especially hydrolysates by Neutrase could
Production of protease by Bacillus amyloliquefaciens in solid-state fermentation and its application in the unhairing of hides and skins
George, S., et al.
Process. Biochem., 30, 457-462 (1995)
R Kammoun et al.
Bioresource technology, 90(3), 249-254 (2003-10-25)
Neutrase, used for hydrolysis of wheat proteins, was inhibited by end-product in a competitive uncompetitive way. The inhibition ratio depends on the progress of protein hydrolysis (degree of hydrolysis) and it remains constant beyond a degree of hydrolysis of 7.5%.
Toshifumi Miyazawa et al.
Protein and peptide letters, 15(10), 1050-1054 (2008-12-17)
Bacillus amyloliquefaciens protease-catalyzed peptide bond formation was investigated using the carbamoylmethyl (Cam) ester as the acyl donor. A series of N-protected L-amino acid Cam esters were coupled with an L-amino acid amide in acetonitrile in good to excellent yields. The
Mariem Guidara et al.
Carbohydrate polymers, 253, 117283-117283 (2020-12-07)
The effect of extraction procedures on chemical composition, structural, antitumor and anticoagulant properties of the sulphated polysaccharide 'ulvan' from the green seaweed Ulva lactuca were investigated. The structural features of ulvans were carried out by FTIR and by one- and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service