Skip to Content
Merck
All Photos(2)

Key Documents

P0029

Sigma-Aldrich

Protease from Bacillus sp.

Synonym(s):

Protamex®

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
3.4.21.14
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Bacillus sp.

Quality Level

form

granular

enzyme activity

>1.5 AU-N/g

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protease from Bacillus sp. is extensively useful in pharmaceutical, leather, food and waste processing industry. It is widely used in manufacturing protein hydrolysates.[1] Analysis of Bacillus protease stability and activity showed that the enzyme was active at all pH values from 5.0-11.0 with 100% activity at 8.0. Optimal pH for protein stability was determined to be at 7.0 while the optimal temperature for stability was at 60°C with 95% activity maintained.[2]

Application

Protease from Bacillus sp. has been used:
  • to evaluate protease inhibitor activity[3]
  • in high yields in batch fermentation using a response surface methodology[4]
  • in a study to investigate a biosensor to detect a protease biomarker from Bacillus licheniformis[5]

Analysis Note

minimum activity 1.5 AU-N/G solid

Legal Information

Protamex is a registered trademark of Novozymes Corp.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of protein hydrolysates and lipids obtained from black scabbardfish (Aphanopus carbo) by-products and antioxidative activity of the hydrolysates produced.
Batista, I., et al
Process. Biochem., 45(1), 18-24 (2010)
Sumant Puri et al.
Current microbiology, 44(4), 286-290 (2002-03-23)
High yields (1939 U/ml) of an alkaline protease were obtained in batch fermentation of a Bacillus sp. using a response surface methodology. The interaction of four variables, viz., starch, peptone, incubation time, and inoculum density, suggested inoculum density to be
Production and properties of an extracellular protease from thermophilic Bacillus sp
<BIG>Nascimentol W, et al.</BIG>
Brazilian Journal of Microbiology, 35, 91-96 (2004)
Some properties of extracellular protease from Bacillus licheniformis LBBL-11 isolated from ?iru?, a traditionally fermented African locust bean condiment.
Olajuyigbe, FM.
African Journal of Biochemistry Research, 2, 206-210 (2008)
Hypocholesterolemic effect of compounded freshwater clam protein hydrolysate and Gracilaria
Lin, Y-H, et al
Food Chemistry, 123(2), 395-399 (2010)

Questions

1–2 of 2 Questions  
  1. Podrían indicarme las temperaturas y pH óptimos de trabajo de esta proteasa

    1 answer
    1. As mentioned in the 'DESCRIPTION' section, the optimal pH for protein stability was determined to be at 7.0 while the optimal temperature for stability was at 60°C with 95% activity maintained.

      Helpful?

  2. What is the composition (or protein content) of this protease?

    1 answer
    1. This is a product of Novozymes. The composition is considered proprietary, however, it is a blend of aggressive endo-proteases.

      See the link below to a publication that may be helpful:
      https://www.sciencedirect.com/science/article/pii/S2772502222000683

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service