Skip to Content
Merck
All Photos(4)

Key Documents

P0378

Sigma-Aldrich

Phosphocholine chloride calcium salt tetrahydrate

Sigma Grade

Synonym(s):

Calcium phosphorylcholine chloride, Choline chloride phosphate calcium salt, Choline phosphate chloride, Phosphorocholine chloride calcium salt, Phosphorylcholine chloride calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO3POCH2CH2N(Cl)(CH3)3 · 4H2O
CAS Number:
Molecular Weight:
329.73
Beilstein:
4835230
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.83

biological source

synthetic (organic)

Quality Level

grade

Sigma Grade

Assay

≥97% anhydrous basis (HCl, titration)

form

powder

solubility

water: 100 mg/mL, clear to hazy, colorless to faintly yellow

functional group

chloro

lipid type

phospholipids

shipped in

ambient

storage temp.

−20°C

SMILES string

O.O.O.O.[Cl-].[Ca++].C[N+](C)(C)CCOP([O-])([O-])=O

InChI

1S/C5H14NO4P.Ca.ClH.4H2O/c1-6(2,3)4-5-10-11(7,8)9;;;;;;/h4-5H2,1-3H3,(H-,7,8,9);;1H;4*1H2/q;+2;;;;;/p-2

InChI key

JGULOZKZZMBGFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Phosphocholine chloride calcium salt tetrahydrate has been used:
  • for the purification of anti-phosphorylcholine (PC)
  • for the hydrolysis of phosphomonoesters
  • for the preparation of a mixed solution of creatine, N-acetylaspartate and phosphocholine to perform perchloric acid (PCA) extraction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recovery Correction Technique for NMR Spectroscopy of Perchloric Acid Extracts using DL-valine-2, 3-d2: Validation and Application to 5-Fluorouracil-induced Brain Damage
Nakagami R, et al.
Magnetic Resonance in Medical Sciences : MRMS : An Official Journal of Japan Society of Magnetic Resonance in Medicine, 13(3), 145-153 (2014)
Mirco Eckardt et al.
ChemistryOpen, 9(11), 1214-1220 (2020-12-10)
Applications in biomedicine and ferrofluids, for instance, require long-term colloidally stable, concentrated aqueous dispersions of magnetic, biocompatible nanoparticles. Iron oxide and related spinel ferrite nanoparticles stabilized with organic molecules allow fine-tuning of magnetic properties via cation substitution and water-dispersibility. Here
Muhammad S Riaz et al.
Frontiers in microbiology, 9, 1903-1903 (2018-09-28)
Mycobacterium tuberculosis, the causative agent of tuberculosis, is one of the leading causes of human deaths due to a single infectious agent. M. tuberculosis infection of the host initiates a local inflammatory response, resulting in the production of a range
Erin D Lewis et al.
International journal of food sciences and nutrition, 66(7), 743-748 (2015-09-25)
Epidemiological studies identify meat as a major source of choline; however, the most comprehensive reference for food choline content, the United States Department of Agriculture (USDA) database for dietary choline, does not include values for meats of importance in some
Phosphorylcholine-dependent cross-reactivity between dental plaque bacteria and oxidized low-density lipoproteins
Schenkein HA, et al.
Infection and Immunity, 69(11), 6612-6617 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service