Skip to Content
Merck
All Photos(2)

Key Documents

P0358

Sigma-Aldrich

Anti-Potassium Channel Kv4.3 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BRGDA9, Anti-KCND3L, Anti-KCND3S, Anti-KSHIVB, Anti-KV4.3, Anti-SCA19, Anti-SCA22

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCND3(3752)
rat ... Kcnd3(65195)

General description

Potassium voltage-gated channel subfamily D member 3 (KCND3) gene codes for Kv4.3, an α-subunit of the Ito channel. It has seven exons and is mapped to the human chromosome 1p13.2. Kv4.3 is expressed in the brain and the heart. Anti-potassium channel Kv4.3 recognizes Kv4.3 protein in the rat by immunoblotting.

Immunogen

synthetic peptide corresponding to amino acids 451-468 of rat or human potassium channel Kv4.3 (with additional N-terminal tyrosine and methionine replaced with norleucine).

Application

Anti-Potassium Channel Kv4.3 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Potassium channels contribute to maintaining cell volume, neuronal excitability, and the secretion of transmitters, salt, and hormones. Potassium voltage-gated channel subfamily D member (Kv4.3) is involved in membrane repolarization in excitable cells. Kv4.3 may participate in the development of the cerebellum. It is capable of forming homo- or hetero-tetramers with the Shal subfamily channel members. KCND3 gene mutations result in Brugada syndrome and spinocerebellar ataxia type 22.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Hyung Cho et al.
JCI insight, 3(19) (2018-10-05)
Sudden death is the most common mode of exodus in patients with heart failure and preserved ejection fraction (HFpEF). Cardiosphere-derived cells (CDCs) reduce inflammation and fibrosis in a rat model of HFpEF, improving diastolic function and prolonging survival. We tested
Saïd F A Askar et al.
Circulation. Arrhythmia and electrophysiology, 6(2), 380-391 (2013-02-20)
After intramyocardial injection, mesenchymal stem cells (MSCs) may engraft and influence host myocardium. However, engraftment rate and pattern of distribution are difficult to control in vivo, hampering assessment of potential adverse effects. In this study, the role of the engraftment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service