Skip to Content
Merck
All Photos(1)

Key Documents

11647229001

Roche

Cell Proliferation ELISA, BrdU (colorimetric)

sufficient for ≤1,000 tests

Synonym(s):

cell proliferation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

usage

sufficient for ≤1,000 tests

Quality Level

manufacturer/tradename

Roche

technique(s)

ELISA: suitable

detection method

colorimetric

storage temp.

2-8°C

General description

Colorimetric immunoassay for the quantification of cell proliferation, based on the measurement of BrdU incorporation during DNA synthesis:  a nonradioactive alternative to the [3H]-thymidine incorporation assay.

Specificity

The antibody conjugate reacts with the thymidine analogue 5-bromo-2′-deoxyuridine (BrdU) and with BrdU incorporated into DNA. For binding to BrdU incorporated into the DNA, the BrdU-labeled DNA has to be denatured. The antibody does not cross-react with any endogenous cellular components such as thymidine, uridine, or DNA.

Application

For research use only. Not for use in diagnostic procedures.
The Cell Proliferation ELISA, BrdU (colorimetric) belongs to the second, improved generation of kits for measuring DNA synthesis. It is a precise, fast, and simple colorimetric alternative to quantitate cell proliferation based on the measurement of BrdU incorporation during DNA synthesis in replicating (cycling) cells. Thus, the Cell Proliferation ELISA can be used in many different in vitro cell systems. For example:
  • Detection and quantification of cell proliferation induced by growth factors and cytokines
  • Determination of the inhibitory or stimulatory effects of various compounds on cell proliferation in environmental and biomedical research, and in the food, cosmetic, and pharmaceutical industries
  • Measurement of the immunoreactivity of lymphocytes, stimulated by mitogens or antigens
  • Analysis of the chemosensitivity of tumor cells to different cytostatic drugs in medical research
  • Testing of biocompatibility of various scaffolds, employed in bone tissue engineering, for bone cell growth

Packaging

1 kit containing 6 components.

Preparation Note

Working concentration: The kit antibody (Anti-BrdU-peroxidase) has, after reconstitution, a concentration of 7.5 U/ml, after dilution 0.075 U/ml. Instead of the kit antibody, you can also use the Anti-BrdU-Antibody, Fab fragments.This antibody is double-concentrated, so you have to dilute it after reconstitution in 1 ml with 1:200.
Working solution: BrdU labeling solution

Dilute BrdU labeling reagent 1:100 with sterile culture medium (resulting concentration: 100 μM BrdU).
For one 96-well MP, 1 ml BrdU labeling solution is required if the cells were cultured in 100 μl /well (10 μl/well) and 2 ml BrdU labeling solution is required if the cells were cultured in 200 μl/well (20 μl/well).

Anti-BrdU-peroxidase stock solution

Dissolve Anti-BrdU-peroxidase in 1.1 ml double-dist. water for 10 minutes and mix thoroughly.

Anti-BrdU-peroxidase working solution

Dilute Anti-BrdU-peroxidase stock solution 1:100 with antibody dilution solution. For one 96-well MP dilute 100 μl Anti-BrdU-peroxidase stock solution in 10 ml antibody dilution solution

Washing solution

Dilute Washing buffer concentrate 1:10 with double-dist. water.
For one 96-well MP dilute 10 ml Washing buffer concentrate with 90 ml double-dist. water.
Storage conditions (working solution): BrdU labeling solution
The undiluted BrdU labeling reagent (1000x): At 2 to 8 °C for several months protected from light.
The diluted BrdU labeling reagent: At 2 to 8 °C stable for several weeks. Store protected from light. For long-term storage it is recommended to store the BrdU labeling solution in aliquots at -15 to -25 °C.
Anti-BrdU-peroxidase stock solution
At 2 to 8 °C for several months. For long-term storage it is recommended to store the solution in aliquots at -15 to -25 °C.
Anti-BrdU-peroxidase working solution
Prepare shortly before use. Do not store.
Washing solution
At 2 to 8 °C for several weeks.

Reconstitution

The working solution for the antibody should be phosphate buffered saline containing 1% BSA, pH 7.4

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • BrdU Labeling Reagent

  • FixDenat ready-to-use

  • Anti-BrdU-peroxidase antibody

  • Antibody Dilution Solution ready-to-use

  • Washing Buffer PBS 10x concentrated

  • Substrate Solution TMB ready-to-use

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Ang Shi et al.
International journal of molecular medicine, 34(1), 237-243 (2014-04-24)
It is known that microRNA-219 (miR-219) expression is downregulated in medulloblastoma. In the present study, we investigated the expression, targets and functional effects of miR-219 in D283-MED medulloblastoma cells. We first demonstrated that miR-219 not only inhibits proliferation, but also
Hongzhi Ma et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6103-6112 (2015-04-29)
Zinc finger protein, X-linked (ZFX) is a transcriptional factor involved in many physiological processes such as embryonic stem cell survival and self-renewal. Though ZFX dysfunctions have been identified in variant human diseases and especially in cancers, its pathological roles have
María Carolina Durán et al.
Journal of nanobiotechnology, 9, 47-47 (2011-10-22)
Reproducibly high transfection rates with low methodology-induced cytotoxic side effects are essential to attain the required effect on targeted cells when exogenous DNA is transfected. Different approaches and modifications such as the use of nanoparticles (NPs) are being evaluated to
Zsuzsanna E Toth et al.
Blood, 111(12), 5544-5552 (2008-02-13)
Granulocyte colony-stimulating factor (G-CSF) induces proliferation of bone marrow-derived cells. G-CSF is neuroprotective after experimental brain injury, but the mechanisms involved remain unclear. Stem cell factor (SCF) is a cytokine important for the survival and differentiation of hematopoietic stem cells.
Christa Broholm et al.
Journal of applied physiology (Bethesda, Md. : 1985), 111(1), 251-259 (2011-04-30)
The cytokine leukemia inhibitory factor (LIF) is expressed by skeletal muscle and induces proliferation of myoblasts. We hypothesized that LIF is a contraction-induced myokine functioning in an autocrine fashion to activate gene regulation of human muscle satellite cell proliferation. Skeletal

Articles

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

See All

Protocols

Cell Proliferation ELISA, BrdU (colorimetric) Protocol & Troubleshooting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service