NA9604
GenElute™ HP 96-Well Plasmid Miniprep Kit
sufficient for 4 96-well plate purifications
Synonym(s):
96 well plasmid extraction kit, 96 well plasmid isolation kit, 96 well plasmid purification kit, High Throughput Purification, Plasmid Purification, high throughput miniprep kit, high throughput plasmid kit, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, Gen Elute, GenElute™ HP Plasmid Kit
About This Item
Recommended Products
usage
sufficient for 4 96-well plate purifications
technique(s)
DNA purification: suitable
storage temp.
15-25°C
Related Categories
General description
The GenElute HP 96 Well Plasmid Miniprep Kit offers a simple, rapid, costeffective solution for high-throughput purification of plasmid DNA from recombinant E. coli cultures. By combining silica-binding technology and the convenience of a vacuum format, up to 10 μg of high copy plasmid DNA per well can be recovered from 1.3 mL of culture in less than 50 minutes. Actual yields and the optimum volume of culture depend on the plasmid and culture medium used.
The purified plasmid DNA is suitable for a wide variety of molecular biology applications, including restriction digestions and sequencing.
Application
Features and Benefits
- Up to 10 μg of high-copy plasmid DNA per well
- Flexibility of both manual and automated formats
- Optimized for production of virus titers from MISSION shRNA plasmid vectors
- No phenol/chloroform extraction or alcohol precipitation required
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
Flash Point(F)
77.0 °F
Flash Point(C)
25 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service