Skip to Content
Merck
All Photos(7)

Key Documents

NA0200

Sigma-Aldrich

GenElute HP Plasmid Midiprep Kit

greener alternative

sufficient for 25 purifications

Synonym(s):

GenElute HP Plasmid Midiprep Kit, HP midiprep kit, fast midiprep kit, fast plasmid isolation kit, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, ultrafast plasmid isolation kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 25 purifications

Quality Level

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA purification: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

GenElute HP Plasmid Midiprep Kit has been used to isolate and purify plasmids.

Features and Benefits

  • From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
  • Up to 350 μg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • No phenol/chloroform extraction or alcohol precipitation required
  • Contains fewer plastic components, reducing the amount of waste

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa E Gray et al.
G3 (Bethesda, Md.), 9(11), 3683-3689 (2019-09-29)
Despite the importance of Aβ aggregation in Alzheimer's disease etiology, our understanding of the sequence determinants of aggregation is sparse and largely derived from in vitro studies. For example, in vitro proline and alanine scanning mutagenesis of Aβ40 proposed core
Genome-wide CRISPR screens reveal a Wnt-FZD5 signaling circuit as a druggable vulnerability of RNF43-mutant pancreatic tumors.
Steinhart Z
Nature Medicine, 23(1), 60-68 (2017)
The distal upstream promoter in Ly49 genes, Pro1, is active in mature NK cells and T cells, does not require TATA boxes, and displays enhancer activity.
Gays F
Journal of Immunology, 194(12), 6068-6081 (2015)
Stephanie P Cartwright et al.
BMC research notes, 10(1), 340-340 (2017-07-29)
Patients with dual hepatitis B (HBV) and hepatitis D (HDV) virus infection are at an increased risk of progression to liver cirrhosis and hepatocellular carcinoma than patients with a single viral infection. Treatment of viral hepatitis due to dual HBV/HDV
Melissa A Chiasson et al.
eLife, 9 (2020-09-02)
Vitamin K epoxide reductase (VKOR) drives the vitamin K cycle, activating vitamin K-dependent blood clotting factors. VKOR is also the target of the widely used anticoagulant drug, warfarin. Despite VKOR's pivotal role in coagulation, its structure and active site remain

Related Content

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service