MAK531
Catalase Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Catalase Activity Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
colorimetric
fluorometric
relevant disease(s)
endocrinological disorders, diabetes; neurological disorders; dermatological diseases; cardiovascular diseases
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Catalase is an ubiquitous antioxidant enzyme that catalyzes the decomposition of hydrogen peroxide (H2O2) to water and oxygen. By preventing excessive H2O2 build up, catalase allows important cellular processes which produce H2O2 as a byproduct to occur safely. Deficiency in catalase activity has been associated with grey hair and peroxisomal disorder acatalasia.
Features and Benefits
- Linear detection range: 0.2 to 5 U/L
Suitability
The kit is suitable for the quantitative determination of catalase activity in serum, plasma, urine, saliva, cell culture etc., as well as for studying the effects of drugs on catalase activity.
Principle
The Catalase Assay Kit directly measures catalase degradation of H2O2 using a redox dye. The change in color intensity at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the catalase activity in the sample.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service