Skip to Content
Merck
All Photos(1)

Key Documents

N8271

Sigma-Aldrich

α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

Neuraminidase from Arthrobacter ureafaciens, Acyl-neuraminyl Hydrolase, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥135 units/mg protein

mol wt

88 kDa
95 kDa

foreign activity

β-Galactosidase, α-mannosidase, β-hexosaminidase, α-fucosidase, and proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Releases α(2→3), α(2→6), α(2→8), and α(2→9)-linked N-acetylneuraminic acid from complex oligosaccharides.

Packaging

Provided with 5× reaction buffer (250 mM sodium phosphate, pH 6.0).

Unit Definition

One unit will hydrolyze 1 μmole of 4-methylumbelliferyl α-D-N-acetylneuraminide per min at pH 5.0 at 37 °C.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, and 20 mM NaCl.

Preparation Note

Expressed in glycosidase-free hosts.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Nordholm et al.
The Journal of biological chemistry, 288(15), 10652-10660 (2013-03-01)
Interactions that facilitate transmembrane domain (TMD) dimerization have been identified mainly using synthetic TMDs. Here, we investigated how inherent properties within natural TMDs modulate their interaction strength by exploiting the sequence variation in the nine neuraminidase subtypes (N1-N9) and the
Dominic Meusch et al.
Nature, 508(7494), 61-65 (2014-02-28)
Tripartite Tc toxin complexes of bacterial pathogens perforate the host membrane and translocate toxic enzymes into the host cell, including in humans. The underlying mechanism is complex but poorly understood. Here we report the first, to our knowledge, high-resolution structures
Audu J Natala et al.
Journal of medical entomology, 50(1), 85-93 (2013-02-23)
Amblyomma variegatum F. are obligate hematophagous ectoparasites of livestock that serve as the vectors of Ehrlichia ruminantium (formerly known as Cowdria ruminantium), the causative agent of heartwater disease. In the light of the fact that they are blood-feeding, their salivary
Jin-Hyo Kim et al.
Science (New York, N.Y.), 340(6128), 71-75 (2013-02-23)
Influenza antiviral agents play important roles in modulating disease severity and in controlling pandemics while vaccines are prepared, but the development of resistance to agents like the commonly used neuraminidase inhibitor oseltamivir may limit their future utility. We report here
Charng-Sheng Tsai et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(7), 2466-2471 (2013-01-30)
Alkyne-hinged 3-fluorosialyl fluoride (DFSA) containing an alkyne group was shown to be a mechanism-based target-specific irreversible inhibitor of sialidases. The ester-protected analog DFSA (PDFSA) is a membrane-permeable precursor of DFSA designed to be used in living cells, and it was

Articles

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Learn about O-linked glycan strategies, O-glycosidase actions, how to remove sialic acid residues, β-Elimination, and O-glycan modifications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service