Merck
  • Identification and characterization of sialidase-like activity in the developmental stages of Amblyomma variegatum.
Identification and characterization of sialidase-like activity in the developmental stages of Amblyomma variegatum.
Journal of medical entomology (2013-02-23)
Audu J Natala, Emmanuel O Balogun, Joshua A B Balogun, Hajiya M Inuwa, Andrew J Nok, Tomoo Shiba, Shigeharu Harada, Kiyoshi Kita, Rowland I S Agbede, King A N Esievo
ABSTRACT
Amblyomma variegatum F. are obligate hematophagous ectoparasites of livestock that serve as the vectors of Ehrlichia ruminantium (formerly known as Cowdria ruminantium), the causative agent of heartwater disease. In the light of the fact that they are blood-feeding, their salivary glands play prominent role in their acquisition of nutrients from the bloodmeal. Sialic acids are a major component of glycoprotein in mammalian blood fluid and cells. Sialome of hard ticks is still sparse. Here, for the first time, the possible expression of sialidase in A. variegatum was investigated. Our finding established the presence of type II sialidase-like activity in the three stages (larva, nymph, and adult) of the fed and unfed tick. There was no statistically significant difference in sialidase activity in the various stages of this ectoparasite (P > 0.05). The enzyme was purified by combination of salting out and ion exchange chromatography on DEAE--cellulose and hydroxylapatite columns. Characterization of the enzyme revealed that it is optimally active at 40 degrees C and pH 5.5, and is activated by bivalent cations Zn2+ or Fe2+. The enzyme has a Km of 0.023 mM and Vmax of 0.16 millimol/min with Fetuin as the substrate. To assess the susceptibility of some mammalian cells to the tick sialidase, we prepared erythrocyte ghost cells from different animals, which were incubated with the enzyme. Results revealed that the ruminant cells were better substrates. Our work and findings contribute to the preliminary characterization of the A. variegatum salivary proteome, and may pave way to the development of new acaricides.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS