Skip to Content
Merck
All Photos(1)

Key Documents

N5538

Sigma-Aldrich

Monoclonal Anti-3-Nitrotyrosine antibody produced in mouse

clone 18G4, purified immunoglobulin, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

18G4, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~55 kDa

species reactivity

wide range

technique(s)

western blot: 1:1,000 using mouse and rat brain

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

nitration (Tyr)

Gene Information

human ... NOS1(4842)

Looking for similar products? Visit Product Comparison Guide

General description

3-nitrotyrosine (3-NT) is synthesized by the incorporation of nitro triatomic group (-NO2) at the third position of the phenolic ring in the tyrosine residue, in response to the production of peroxynitrite in the oxidative and nitrosative stress.

Immunogen

3-Nitrotyrosine-KLH.

Application

Monoclonal Anti-3-Nitrotyrosine antibody produced in mouse has been used in western blot analysis and immunohistochemistry.

Biochem/physiol Actions

3-Nitrotyrosine (3-NT) in tissue extracts is a marker for the determination of protein damage induced by peroxynitrite. 3-NT is used as a biomarker to determine the protein nitration, associated with various disorders. It is implicated in diseases characterized by endothelial dysfunction as in stroke, myocardial infarction, atherosclerosis and diabetes.

Target description

Monoclonal Anti-3-Nitrotyrosine recognizes 3-nitrotyrosine in most species. Protein tyrosine nitration results in a post-translational modification that is increasingly receiving attention as an important component of nitric oxide signaling.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiotrophin-1 reduces ischemia/reperfusion injury during liver transplant
Aguilar-Melero P, et al.
The Journal of Surgical Research, 181(2), e83-e91 (2013)
Phytochemical analysis and biological properties of Cyperus rotundus L.
Kumar KH, et al.
Industrial Crops and Products, 52, 815-826 (2014)
Pengcheng Xu et al.
Frontiers in cellular neuroscience, 15, 804345-804345 (2022-01-04)
Mutations in a number of genes encoding mitochondrial aminoacyl-tRNA synthetases lead to non-syndromic and/or syndromic sensorineural hearing loss in humans, while their cellular and physiological pathology in cochlea has rarely been investigated in vivo. In this study, we showed that
Andrographolide Ameliorates Diabetic Cardiomyopathy in Mice by Blockage of Oxidative Damage and NF-kappaB-Mediated Inflammation
Liang E, et al.
Oxidative Medicine and Cellular Longevity, 2018 (2018)
Amanda M Eccardt et al.
Catalysts (Basel, Switzerland), 12(8) (2023-05-01)
Iron porphyrin molecules such as hemin and iron(III) 4,4',4″,4‴-(porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid) (FeTBAP) have previously been shown to influence insulin signaling and glucose metabolism. We undertook this study to determine whether a catalytic action of iron porphyrin compounds would be related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service