Skip to Content
Merck
All Photos(1)

Key Documents

05-233

Sigma-Aldrich

Anti-Nitrotyrosine Antibody, clone 1A6

clone 1A6, Upstate®, from mouse

Synonym(s):

Anti-nitrotyrosine antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A6, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NOS1(4842)

General description

Nitrotyrosine has been identified as an indicator of cell damage and inflammation, as well as of the production of NO. It is believed that measuring the concentration of nitrotyrosine will serve as a marker for damage caused by NO in the cell. Previous studies have shown that nitrotyrosine has been found in inflammatory conditions such as atherosclerotic plaques and rheumatoid arthritis, among others

Application

Anti-Nitrotyrosine Antibody, clone 1A6 detects level of Nitrotyrosine & has been published & validated for use in IC, IH, IP & WB.

Quality

routinely evaluated on peroxynitrite-treated A431 cells; no reaction was noted with non-nitrated proteins

Target description

Predicted MW for nitrated-BSA: 66 kDa

Physical form

Format: Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide, and 30% glycerol.

Analysis Note

Control
Immunohistochemistry: Human Alzheimer disease brain sections (Methacarn fixed), mouse liver
Immunoblotting: Nitrated-BSA
Indirect ELISA: nitrated-BSA
Indirect immunofluorescence: A549 cells metabolically labeled with 3-nitrotyrosine

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The extensive nitration of neurofilament light chain in the hippocampus is associated with the cognitive impairment induced by amyloid beta in mice.
Alkam, T; Nitta, A; Mizoguchi, H; Itoh, A; Murai, R; Nagai, T; Yamada, K; Nabeshima, T
Journal of Pharmacology and Experimental Therapeutics null
Relaxin ameliorates hypertension and increases nitric oxide metabolite excretion in angiotensin II but not N(?)-nitro-L-arginine methyl ester hypertensive rats.
Sasser, JM; Molnar, M; Baylis, C
Hypertension null
Microparticles from apoptotic monocytes enhance nitrosative stress in human endothelial cells.
Mastronardi, Maria Letizia, et al.
Fundamental & Clinical Pharmacology, 25, 653-660 (2011)
Peroxynitrite diminishes myogenic activity and is associated with decreased vascular smooth muscle F-actin in rat posterior cerebral arteries.
Maneen, MJ; Hannah, R; Vitullo, L; DeLance, N; Cipolla, MJ
Stroke null
Han Wang et al.
Cardiovascular diabetology, 16(1), 12-12 (2017-01-21)
Endothelial dysfunction plays a critical role in the development of type 2 diabetes (T2DM). T-cadherin (T-cad) has gained recognition as a regulator of endothelial cell (EC) function. The present study examined whether T-cad deficiency increases vascular vulnerability in T2DM. Vascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service