MSKT0004
IA-MS Immunoaffinity Mass Spectrometry Kit - Protein A/G
96 Well Strip Plate with SOLu-Trypsin
Synonym(s):
Target protein enrichment and quantification from complex background by mass spectrometry.
About This Item
Recommended Products
form
solid
suitability
suitable for mass spectrometry
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
Application
Features and Benefits
- Robust, high-throughput quantification of proteins in complex backgrounds by LC-MS/MS
- Entire workflow including digest completed in under 5 hours, such that samples are injection ready at the end of the work day
- 96-well plate format has potential for automation in a liquid-handling system
- Bead-free system circumvents messy clean-up and eliminates risk of column clogging
- Plate strips allow user to run as many or as few samples as desired
Packaging
Components
SOLu-Trypsin EMS0004-100UG
MS Denaturation Solution EMS00010-10ML
Rapid Trypsin Digestion Buffer EMS0009-30ML
Tris Buffered Saline with TWEEN® 20 (TBST)powder, pH 8.0 T9039
Tris Buffered Saline (TBS) powder, pH 8.0 T6664
EZ-Pierce™ plate seal Z721581-1EA
See datasheet for more information.
Storage and Stability
Related Presentation
Legal Information
Kit Components Only
- MS Denaturation Solution
- Rapid Trypsin Digestion Buffer
Kit Components Also Available Separately
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
Flash Point(F)
75.2 °F
Flash Point(C)
24 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service