Skip to Content
Merck
All Photos(1)

Key Documents

MITOISO3

Sigma-Aldrich

Yeast Mitochondria Isolation Kit

sufficient for 40 applications (using 20 OD culture preparations), isolation of an enriched mitochondrial fraction from yeast cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

usage

sufficient for 40 applications (using 20 OD culture preparations)

storage temp.

−20°C

General description

Mitochondria, the site of most of the energy production in eukaryotic cells, are characterized by a double membrane structure, an outer membrane and folded inner membrane. Isolated mitochondria may be used as tools for in vitro studies such as respiration and energy production studies, apoptosis, mtDNA and mtRNA, and mitochondrial protein profiling.

Application

The kit contains all the reagents required for yeast cell wall lysis by lyticase (spheroplast formation) followed by cell membrane lysis/breakage and mitochondria isolation. In addition, the kit includes an extraction buffer for mitochondrial protein profiling to be used in proteome studies and a storage buffer for use with intact mitochondria. Yeast Mitochondria Isolation Kit has been used in the isolation and extraction of mitochondria.

Features and Benefits

• The kit′s features include speed, sensitivity, ease of use, and specificity
• The kit contains all the reagents required for a fast and easy yeast cell wall lysis by lyticase (spheroplast formation) followed by cell membrane lysis/breakage and mitochondria isolation
• The kit provides two alternatives for cell membrane disruption. The first one by a detergent and second one by homogenization.
• The kit includes an extraction buffer for mitochondrial protein profiling to be used in proteome studies
• The kit includes a storage buffer for use with intact mitochondria.
• The reagents are sufficient for 40 procedures using 20 OD culture preparations
• The kit was tested on Saccharomyces cerevisiae, Pichia pastoris, and Schizosaccharomyces pombe

Kit Components Also Available Separately

Product No.
Description
SDS

  • L2524Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuretSDS

  • P8215Protease Inhibitor Cocktail, for use with fungal and yeast extracts, DMSO solutionSDS

  • C0356Protein Extraction Reagent Type 4SDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maria Guirola et al.
Metallomics : integrated biometal science, 6(3), 634-645 (2014-02-08)
Zinc is an essential metal for all organisms, as it participates in the structure and/or function of many proteins. However, zinc excess is as deleterious to cells as zinc deficiency. A genome-wide study of the transcriptomic response to high zinc
Yao Wan et al.
Genes & development, 32(19-20), 1309-1314 (2018-09-20)
The mitochondrial cytoplasmic surface serves as a processing site for numerous RNAs from budding yeast to metazoans. We report that budding yeast mitochondrial outer membrane (MOM) proteins that are subunits of the translocase of the outer mitochondrial membrane (Tom70 and
Erika Steele et al.
Microbial cell factories, 20(1), 138-138 (2021-07-21)
Myo-Inositol Phosphate Synthase (MIP) catalyzes the conversion of glucose 6- phosphate into inositol phosphate, an essential nutrient and cell signaling molecule. Data obtained, first in bovine brain and later in plants, established MIP expression in organelles and in extracellular environments.
María Guirola et al.
The Biochemical journal, 432(3), 595-605 (2010-09-23)
The Saccharomyces cerevisiae gene PIF1 encodes a conserved eukaryotic DNA helicase required for both mitochondrial and nuclear DNA integrity. Our previous work revealed that a pif1Δ strain is tolerant to zinc overload. In the present study we demonstrate that this
Christian Streng et al.
The EMBO journal, 40(17), e108083-e108083 (2021-07-14)
Mitochondria are essential organelles because of their function in energy conservation. Here, we show an involvement of mitochondria in phytochrome-dependent light sensing in fungi. Phytochrome photoreceptors are found in plants, bacteria, and fungi and contain a linear, heme-derived tetrapyrrole as

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service