Skip to Content
Merck
All Photos(1)

Key Documents

MIT1000

Sigma-Aldrich

Mitochondria/Cytosol Fractionation Kit

This Mitochondria/Cytosol Fractionation Kit provides reagents for quick & efficient isolation of intact mitochondria from cultured cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.75

Quality Level

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

application(s)

sample preparation

shipped in

dry ice

General description

Mitochondria, sometimes described as the “power plants” of the cell, are sites where most of the energy production in eukaryotic cells takes place. The synthesis of most of the adenosine triphosphate (ATP) occurs in these double membraned organelles that are found in living cells. ATP production by the mitochondria is done by the process of respiration, which uses oxygen to generate energy. This is a very efficient process for using food energy to make ATP. In addition to supplying cellular energy, mitochondria are involved in a range of other processes, such as signaling, cellular differentiation, cell death, as well as the control of the cell cycle and cell growth. Mitochondria also regulate crucial apoptosis signaling pathways. The number of mitochondria in a cell varies widely by organism and tissue type. Many cells have only a few mitochondria, whereas others can contain several thousand.

Millipore’s Mitochondria/Cytosol Fractionation Kit provides reagents for quick and efficient isolation of intact mitochondria from cultured cells. This kit allows mitochondrial isolation by using a convenient table top microcentrifuge, and can be used to separate an enriched mitochondrial fraction (heavy membrane fraction) from cytosolic fraction (light membrane fraction). Such separation is useful for studying apoptosis and signaling pathways between the two fractions, by Western blotting or ELISA.

The kit contains sufficient materials for the analysis of 50 samples.

Application

Research Category
Apoptosis & Cancer

Components

Isotonic Mitochondrial Buffer (CS204255): 50 mL

Mitochondrial Lysis Buffer (CS204257): 5 mL

Protease inhibitor cocktail (CS204253): 1 mL

Anti-Bcl2 (05-729-25UG): 25 μg

Anti-GAPDH (CS204254): 25 μg

Storage and Stability

All components should be store at -20°C for up to one year from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongmei Li et al.
Nature cell biology, 15(7), 773-785 (2013-06-25)
Following exocytosis, the rate of recovery of neurotransmitter release is determined by vesicle retrieval from the plasma membrane and by recruitment of vesicles from reserve pools within the synapse, which is dependent on mitochondrial ATP. The anti-apoptotic Bcl-2 family protein
Jinxin Pan et al.
Central-European journal of immunology, 47(1), 20-29 (2022-05-24)
The protective activity of scopoletin (SPT) against glucose-induced cataract has been attributed to attenuation of aldose reductase activity and oxidative stress in a rat model. The present investigation was aimed to study the protective effect and mechanism of SPT in
Simon C Baker et al.
The American journal of pathology, 186(5), 1267-1277 (2016-03-24)
Recreational abuse of ketamine has been associated with the emergence of a new bladder pain syndrome, ketamine-induced cystitis, characterized by chronic inflammation and urothelial ulceration. We investigated the direct effects of ketamine on normal human urothelium maintained in organ culture
Turki Y Alhazzazi et al.
Anticancer research, 36(1), 49-60 (2016-01-02)
The survival rate of patients with head and neck squamous cell carcinoma (HNSCC) stands at approximately 50% and this has not improved in decades. This study developed a novel sirtuin-3 (SIRT3) inhibitor (LC-0296) and examined its role in altering HNSCC
Yoshimi Endo Greer et al.
Oncotarget, 9(26), 18454-18479 (2018-05-03)
We report a novel mechanism of action of ONC201 as a mitochondria-targeting drug in cancer cells. ONC201 was originally identified as a small molecule that induces transcription of TNF-related apoptosis-inducing ligand (TRAIL) and subsequently kills cancer cells by activating TRAIL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service