Skip to Content
Merck
All Photos(2)

Key Documents

MAK190

Sigma-Aldrich

ATP Assay Kit

Sufficient for 100 Colorimetric or Fluorometric tests

Synonym(s):

Adenosine triphosphate Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric
fluorometric

relevant disease(s)

cancer; neurological disorders

storage temp.

−20°C

General description

Adenosine triphosphate (ATP) is a nucleoside triphosphate formed exclusively in the mitochondria. It is a high-energy molecule described as the energy currency in all living systems. The chemical energy contained in the phosphate bond of ATP drives most cellular processes. Genetic disorders such as Leber hereditary optic neuropathy, Leigh syndrome, neuropathy, ataxia, and retinitis pigmentosa affect the generation of ATP in the mitochondria.

Application

ATP Assay Kit has been used to quantify the ATP concentration by fluorometric detection of ATP in samples.

Suitability

Suitable for the determination of ATP in samples such as animal tissue and cell culture samples.

Principle

The ATP Colorimetric/Fluorometric Assay Kit is a sensitive assay for determining ATP in various samples (ranging from 2–10 nmole/well for the colorimetric assay and from 20–1,000 pmole/well for the fluorometric assay). ATP concentration is determined by phosphorylating glycerol, resulting in a colorimetric (570 nm) or fluorometric (lex = 535/lem = 587 nm) product proportional to the amount of ATP present.

Other Notes

The fluorometric assay is 10-100 times more sensitive than the colorimetric assay.

Kit Components Only

Product No.
Description

  • ATP Assay Buffer

  • ATP Probe, in DMSO

  • ATP Converter

  • Developer Mix

  • ATP Standard, 1.0 μmole

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cost of surviving sepsis: A novel model of recovery from sepsis in Drosophila melanogaster.
Kaynar A M, et al.
Intensive care medicine experimental, 4(1), 4-4 (2016)
E Alirol et al.
Oncogene, 25(34), 4706-4716 (2006-08-08)
Mitochondria are key players in several cellular functions including growth, division, energy metabolism, and apoptosis. The mitochondrial network undergoes constant remodelling and these morphological changes are of direct relevance for the role of this organelle in cell physiology. Mitochondrial dysfunction
Melatonin Balance the Autophagy and Apoptosis by Regulating UCP2 in the LPS-Induced Cardiomyopathy.
Pan P, et al.
Molecules (Basel), 23(3), 675-675 (2018)
Brock A Humphries et al.
Breast cancer research : BCR, 22(1), 60-60 (2020-06-07)
Mitochondrial dynamics underlies malignant transformation, cancer progression, and response to treatment. Current research presents conflicting evidence for functions of mitochondrial fission and fusion in tumor progression. Here, we investigated how mitochondrial fission and fusion states regulate underlying processes of cancer
Sandra Aurora Niño et al.
ACS chemical neuroscience, 10(1), 323-336 (2018-08-25)
Worldwide, every year there is an increase in the number of people exposed to inorganic arsenic (iAs) via drinking water. Human populations present impaired cognitive function as a result of prenatal and childhood iAs exposure, while studies in animal models

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service