MAK473
ATP Assay Kit
sufficient for 100 assays (bioluminescent)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
bioluminescent
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Adenosine 5′-triphosphate (ATP) is the chemical energy source for cellular metabolism and is often referred to as “energy currency" of the cell. ATP is produced in living cells during photosynthesis and cellular respiration and consumed in cellular processes including biosynthetic reactions, motility, and cell division. ATP is a key indicator of cellular activity and has been utilized as a measure of cell viability and cytotoxicity in research and drug discovery.
Features and Benefits
- Sensitive and accurate. The kit can detect as low as 0.02 µM ATP, or a single cell can be quantified.
- Compatible with high-throughput handling systems.
- Safe. Non-radioactive assay.
Suitability
The kit is suitable for ATP determination in cells, tissue, and other biological samples.
Principle
The ATP Assay Kit provides a rapid method to measure intracellular ATP. The single working reagent lyses cells to release ATP, which in the presence of luciferase, immediately reacts with the substrate D luciferin to produce light. The light intensity is a direct measure of intracellular ATP.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service