Skip to Content
Merck
All Photos(1)

Key Documents

CGD1

Sigma-Aldrich

Cell Growth Determination Kit, MTT based

Synonym(s):

Mitochondrial activity assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

packaging

pkg of 1 kit

Quality Level

technique(s)

UV/Vis spectroscopy: suitable
protein quantification: suitable

Amax

570 nm

application(s)

cell analysis
detection

detection method

colorimetric

shipped in

dry ice

storage temp.

−20°C

General description

Cell Growth Determination Kit (MTT based) is designed for the spectrophotometric measurement of cell growth as a function of mitochondrial activity in living cells. The MTT (3-(4, 5-dimethylthiazolyl-2)-2, 5-diphenyltetrazolium bromide) system is a simple, accurate, reproducible means of measuring the activity of living cells via mitochondrial dehydrogenase activity.

Application

Cell Growth Determination Kit, MTT based has been used:
  • to measure the cell viability of preosteoblastic cells
  • in MTT (3-(4, 5-dimethylthiazolyl-2)-2, 5-diphenyltetrazolium bromide) assay of normal human prostatic stromal and epithelial cell lines
  • to determine the cell viability of primary cervical epithelial cells

Packaging

Each kit includes 5 vials each containing 1 mL of 5 mg/ml MTT in RPMI (without phenol red) and 50 mL of MTT solvent (0.1 N HCl in anhydrous isopropanol). Volumes of kit components have been optimized for cultures grown in multi-well plates. MTT solutions is added at 10% of total culture volume, thus the number of tests per kit is dependent upon culture vessel and/or volume.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla M R Lacerda et al.
American journal of physiology. Heart and circulatory physiology, 302(10), H1983-H1990 (2012-02-22)
This study addressed the following questions: 1) Does cyclic tensile strain induce protein expression patterns consistent with myxomatous degeneration in mitral valves? 2) Does cyclic strain induce local serotonin synthesis in mitral valves? 3) Are cyclic strain-induced myxomatous protein expression
Qiuyu Lin et al.
Journal of cellular and molecular medicine, 22(10), 4640-4652 (2018-07-25)
This study is aimed to investigate the methylation level of candidate genes and its impact on thyroid carcinoma (THCA) development. Infinium Human Methylation 450 BeadChip Arrays by Illumina (Illumina HM450K) was the most popular CpG microarray platform widely used in
Ahran Pae et al.
The journal of advanced prosthodontics, 6(2), 96-102 (2014-05-21)
This study was performed to characterize the effects of zirconia coated with calcium phosphate and hydroxyapatite compared to smooth zirconia after bone marrow-derived osteoblast culture. Bone marrow-derived osteoblasts were cultured on (1) smooth zirconia, (2) zirconia coated with calcium phosphate
Mengxiong Wang et al.
Oncogene, 38(22), 4264-4282 (2019-02-06)
While HER2 and EGFR are overexpressed in breast cancers and multiple other types of tumors, the use of EGFR and/or HER2 inhibitors have failed to cure many cancer patients, largely because cancers acquire resistance to HER2/EGFR-specific drugs. Cancers that overexpress
Mariarita Perri et al.
Biochimica et biophysica acta, 1800(9), 993-1001 (2010-07-06)
Vitamin A is suggested to be protective against oxidative stress. However, different authors observed pro-oxidant effects of retinoids both in experimental works and clinical trials. These discordances are the bases for the investigation of the proliferative and anti-proliferative properties of

Articles

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service