Skip to Content
Merck
All Photos(1)

Key Documents

M5028

Sigma-Aldrich

Monoclonal Anti-5MTH Folic Acid antibody produced in mouse

clone FA-24, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

FA-24, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:5,000

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-5MTH Folic Acid antibody is specific for an epitope present on 5MTHFA and does not detect folic acid. This hybridoma is a cloned cell line derived from a fusion between a mouse myeloma cell line and splenocytes from BALB/c mice immunized with 5MTH Folic acid conjugated to KLH.

Immunogen

5MTH folic acid, conjugated to KLH.

Application

Monoclonal Anti-5MTH Folic Acid antibody is suitable for use in indirect (Enzyme Linked Immuno Sorbent Assay) ELISA (1:5,000) and competitive ELISA.
Monoclonal Anti-5MTH Folic Acid antibody is suitable for use in indirect ELISA (1:5,000).

Biochem/physiol Actions

5-Methyltetrahydrofolate (5-MTHF) is a cofactor that regulates the conversion of homocysteine to methionine in the presence of another co-factor, vitamin B12. 5-MTHF is also involved in modulating antioxidative and cognitive functions. Increased serum 5-MTHF levels have been linked to anemia and macrocytosis. The antibody reacts with 5MTHFA as a free reagent or when conjugated to carriers such as KLH or BSA. Furthermore, the product associates with 5MTHFA bound to the endogenous folate binder in human plasma and serum. The monoclonal antibody does not cross-react with tetrahydrofolic acid, folinic acid, dihydrofolic acid, and vitamin B12.
Vitamin B12 and folic acid are metabolically interrelated. The enzyme methionine synthetase, which catalyzes the conversion of homocysteine to methionine, requires vitamin B12 and 5MTHFA as cofactors. In the absence of vitamin B12, 5MTHFA cannot be converted to tetra hydro folic acid and enter the metabolic pool of 1-carbon fragment acceptors. Vitamin B12 and folate deficiencies are the most common causes of megaloblastic anemia, abnormal hemopoiesis, interference in the maintenance of normal nerve tissue and general intracellular uptake and function disorders in humans. Folic acid deficiency is common in pregnant women, alcoholics, those whose diets do not include raw fruits and vegetables and people with structural or functional damage to the small intestine.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folic acid deficiency and cancer: mechanisms of DNA instability
Duthie S J.
British Medical Bulletin, 55(3), 578-592 (1999)
Bashir M Rezk et al.
FEBS letters, 555(3), 601-605 (2003-12-17)
The presumed protective effect of folic acid on the pathogenesis of cardiovascular, hematological and neurological diseases and cancer has been associated with the antioxidant activity of folic acid. Peroxynitrite (PON) scavenging activity and inhibition of lipid peroxidation (LPO) of the
Martha Savaria Morris et al.
The American journal of clinical nutrition, 91(6), 1733-1744 (2010-04-02)
Folate deficiency has serious consequences for the fetus. Folic acid fortification of food addresses this problem. However, clinical consequences of vitamin B-12 deficiency may be worsened by high folic acid intakes, perhaps as a direct result of unmetabolized folic acid
Intestinal absorption of folic acid - new physiologic & molecular aspects
Milman, N.
The Indian Journal of Medical Research, 136(5), 725-725 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service