Hep G2 Cell Line human
85011430, human liver, Epithelial
Synonym(s):
HepG2 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
Hep G2 Cell Line human, hepatocyte carcinoma, 85011430
biological source
human liver
growth mode
Adherent
karyotype
Modal no. 55
morphology
Epithelial
products
Prothrombin, antithromin III, alpha-foetoprotein complement, C3 and C4 activator, fibrinogen
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian hepatocyte carcinoma
Cell Line Description
The Hep G2 cell line has been isolated from a liver biopsy of a male Caucasian aged 15 years, with a well differentiated hepatocellular carcinoma. The cells secrete a variety of major plasma proteins e.g. albumin, α2-macroglobulin, α 1-antitrypsin, transferrin and plasminogen. They have been grown successfully in large scale cultivation systems. Hepatitis B virus surface antigens have not been detected. The cells will respond to stimulation with human growth hormone.
Application
Hep G2 cell line human has been used to study the cytotoxicity of carbon monoxide-releasing molecules and organic solvents on HepG2 cells and to evaluate the antiproliferative activity of aqueous extracellular polysaccharides (AEPS) for HepG2 using the MTT colorimetric assay.
Study of hepatocyte function and specific protein expression
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 9,13
D16S539: 12,13
D5S818: 11,12
D7S820: 10
THO1: 9
TPOX: 8,9
vWA: 17
CSF1PO: 10,11
D13S317: 9,13
D16S539: 12,13
D5S818: 11,12
D7S820: 10
THO1: 9
TPOX: 8,9
vWA: 17
Culture Medium
EMEM (EBSS)+ 2mM Glutamine + 1% Non Essential Amino Acids NEAA) + 10% Foetal Bovine Serum FBS / FCS.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Cells might take 6 days to form a confluent monolayer without media change if seeded at 1:4. Until then growth occurs mainly in islands.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service