Skip to Content
Merck
All Photos(3)

Key Documents

M1568

Sigma-Aldrich

Mannose triflate

For PET imaging, ≥98% (TLC)

Synonym(s):

β-D-Mannopyranose 1,3,4,6-tetra-O-acetate 2-O-trifluoromethanesulfonate, 1,3,4,6-Tetra-O-acetyl-2-O-trifluoromethanesulfonyl-β-D-mannopyranose, Mannose triflate, TATM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19F3O12S
CAS Number:
Molecular Weight:
480.36
Beilstein:
4341413
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

PET imaging: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(=O)OC[C@H]1O[C@@H](OC(C)=O)[C@@H](OS(=O)(=O)C(F)(F)F)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C15H19F3O12S/c1-6(19)25-5-10-11(26-7(2)20)12(27-8(3)21)13(14(29-10)28-9(4)22)30-31(23,24)15(16,17)18/h10-14H,5H2,1-4H3/t10-,11-,12+,13+,14-/m1/s1

InChI key

OIBDVHSTOUGZTJ-PEBLQZBPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mannose triflate is a glucose analogue.

Application

Mannose triflate is a well-known precursor for 18F-FDG synthesis for PET applications. The binding of 18F to the mannose triflate has been carried out via SN2 nucleophilic substitution reaction. It is used in computational tomography, an imaging technique in early detection of cancer.

Other Notes

This formulation of mannose triflate has reduced trace impurities that are known to cause lower synthetic yields.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvador Castaneda Vega et al.
NeuroImage, 155, 245-256 (2017-05-06)
The clinical use of Magnetic Resonance Imaging (MRI) and Positron Emission Tomography (PET) has proven to be a strong diagnostic tool in the field of neurology. The reliability of these methods to confirm clinical diagnoses has guided preclinical research to
Martin Thunemann et al.
Nature communications, 8(1), 444-444 (2017-09-07)
Many pathophysiological processes are associated with proliferation, migration or death of distinct cell populations. Monitoring specific cell types and their progeny in a non-invasive, longitudinal and quantitative manner is still challenging. Here we show a novel cell-tracking system that combines
Padgett, H., et al.
Applied Radiation and Isotopes, 40, 433-433 (1989)
Shane B Claggett et al.
EJNMMI research, 3(1), 53-53 (2013-07-17)
Many automated radiosynthesizers for producing positron emission tomography (PET) probes provide a means for the operator to create custom synthesis programs. The programming interfaces are typically designed with the engineer rather than the radiochemist in mind, requiring lengthy programs to
Gulcan Unak et al.
Colloids and surfaces. B, Biointerfaces, 90, 217-226 (2011-11-11)
A new architecture has been designed by the conjugation of [(18)F]2-fluoro-2-deoxy-D-glucose ((18)F-FDG), gold nanoparticles (AuNPs), and anti-metadherin (Anti-MTDH) antibody which is specific to the metadherin (MTDH) over-expressed on the surface of breast cancer cells. Mannose triflate molecule is used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service