Skip to Content
Merck
All Photos(1)

Key Documents

L9919

Sigma-Aldrich

LY2033298

≥98% (HPLC)

Synonym(s):

3-Amino-5-chloro-N-cyclopropyl-6-methoxy-4-methyl-thieno[2,3-b]pyridine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClN3O2S
CAS Number:
Molecular Weight:
311.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

off-white to tan

solubility

DMSO: >20 mg/mL

originator

Eli Lilly

storage temp.

−20°C

SMILES string

COc1nc2sc(C(=O)NC3CC3)c(N)c2c(C)c1Cl

InChI

1S/C13H14ClN3O2S/c1-5-7-9(15)10(11(18)16-6-3-4-6)20-13(7)17-12(19-2)8(5)14/h6H,3-4,15H2,1-2H3,(H,16,18)

InChI key

CTEGQKDJTBWFHW-UHFFFAOYSA-N

Biochem/physiol Actions

LY2033298 is a positive allosteric modulator of muscarinic M4 receptor.
LY2033298 is a robust allosteric potentiator that is highly selective for the human M4 muscarinic acetylcholine receptor subtype. LY2033298 potentiates ACh-M4 binding, with no effect at M1/3/5 receptors. LY2033298 can also bind to the M ACh receptor, and mediate either positive or negative allosteric effects depending on the ligand used to probe receptor activity.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana K Shirey et al.
Nature chemical biology, 4(1), 42-50 (2007-12-07)
Muscarinic acetylcholine receptors (mAChRs) provide viable targets for the treatment of multiple central nervous system disorders. We have used cheminformatics and medicinal chemistry to develop new, highly selective M4 allosteric potentiators. VU10010, the lead compound, potentiates the M4 response to
Vindhya Nawaratne et al.
Molecular pharmacology, 74(4), 1119-1131 (2008-07-17)
The M4 muscarinic acetylcholine (ACh) receptor (mAChR) is a potential therapeutic target but characterized by a lack of subtype-selective ligands. We recently generated "designer receptors exclusively activated by a designer drug" (DREADDs), which contained mutations of two conserved orthosteric-site residues
Jing Wu et al.
European journal of pharmacology, 882, 173274-173274 (2020-06-14)
Opioids strongly inhibit GABAergic neurons in the rostromedial tegmental nucleus (RMTg) that expresses μ-opioid receptors to induce rewarding and psychomotor effects. M3 and M4 muscarinic receptors are co-localized with μ-opioid receptors at these GABAergic neurons. This study explored whether RMTg
W Y Chan et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(31), 10978-10983 (2008-08-06)
Current antipsychotics provide symptomatic relief for patients suffering from schizophrenia and related psychoses; however, their effectiveness is variable and many patients discontinue treatment due to side effects. Although the etiology of schizophrenia is still unclear, a leading hypothesis implicates an
Michael Bubser et al.
ACS chemical neuroscience, 5(10), 920-942 (2014-08-20)
Positive allosteric modulators (PAMs) of the M4 muscarinic acetylcholine receptor (mAChR) represent a novel approach for the treatment of psychotic symptoms associated with schizophrenia and other neuropsychiatric disorders. We recently reported that the selective M4 PAM VU0152100 produced an antipsychotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service