Skip to Content
Merck
All Photos(1)

Key Documents

L7527

Sigma-Aldrich

Lipoprotein, very low density from human plasma

≥95% (SDS-PAGE), solution

Synonym(s):

Pre-β-lipoprotein, VLDL, Very low density lipoprotein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

human plasma

Quality Level

Assay

≥95% (SDS-PAGE)

form

solution

UniProt accession no.

functional group

ester
phospholipid

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

VLDL (Very low density lipoprotein) may be used to study the mechanisms and effects of very low density receptors (VLDLR). VLDL is used as a potential source of fatty acids that support cardiac energy metabolism. VLDL is often studied to differentiate its effects from those of chylomicrons.
Inhibits DNA synthesis in lymphocytes activated by the nonspecific mitogen concanavalin A (Con A).

Physical form

Solution in 150 mM NaCl and 0.01% EDTA, pH 7.4

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akifumi Kushiyama et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(8), 1986-1993 (2013-05-25)
Resistin-like molecule (RELM) β is a secretory protein homologous to resistin and reportedly contributes to local immune response regulation in gut and bronchial epithelial cells. However, we found that activated macrophages also express RELMβ and thus investigated the role of
Gwang-Woong Go et al.
The Yale journal of biology and medicine, 85(1), 19-28 (2012-03-31)
The LDLR family of proteins is involved in lipoproteins trafficking. While the role of LDLR in cardiovascular disease has been widely studied, only recently the role of other members of the LDLR proteins in lipoprotein homeostasis and atherosclerosis has emerged.
Samata Tiwari et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(5), 1079-1086 (2012-04-21)
Steady increase in the incidence of atherosclerosis is becoming a major concern not only in the United States but also in other countries. One of the major risk factors for the development of atherosclerosis is high concentrations of plasma low-density
You-Guo Niu et al.
Journal of lipids, 2011, 189876-189876 (2011-07-21)
The heart is a major consumer of energy and is able to utilise a wide range of substrates including lipids. Nonesterified fatty acids (NEFA) were thought to be a favoured carbon source, but their quantitative contribution is limited because of
M C Sampedro et al.
Biochemical and biophysical research communications, 285(2), 393-399 (2001-07-11)
Human very-low-density lipoprotein (VLDL) inhibits DNA synthesis in lymphocytes activated by the nonspecific mitogen concanavalin A (Con A). We studied the effects of VLDL on lymphocyte activation (IL-2 receptor expression), cell cycle progression, and production of IL-2 and of IL-4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service