Skip to Content
Merck
All Photos(1)

Key Documents

LP1

Sigma-Aldrich

VLDL, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

Assay

>98% (SDS-PAGE)

form

liquid

manufacturer/tradename

Chemicon®

shipped in

wet ice

General description

DENSITY:

0.95-1.006 g/mL
Product Source: Human plasma tested negative for HBsAg and HIV-I, HIV-II, HBc and Hepatitis C antibodies. All blood products should be treated as potentially infectious.
VLDL is isolated from fresh human plasma containing EDTA and sodium citrate by sequential isopycnic ultracentrifugation at the lower and upper density limits of the specified density range (Havel et al., 1955). Density adjustments are performed with solid KBr. VLDL is washed by centrifugation at the upper density limit to ensure removal of higher density proteins.

Quality

QUALITY ASSURANCE:

Protein determination by the Pierce BCA method using 1% SDS with BSA as the standard. Purity of each preparation is verified by agarose gel electrophoresis using Fat red 7B staining for lipid (2,3).

Physical form

Liquid in 150 mM NaCl, pH 7.4 with 0.01% EDTA. Filtered to sterility using 0.2 micron filter in a sterile vial.

COMPOSITION:

88-95% lipid, 5-12% protein

Storage and Stability

Maintain at 2-8°C for up to four weeks. DO NOT FREEZE. Freezing may cause denaturation.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role of Rnd1 GTPase in dendritic spine formation in hippocampal neurons.
Yukio Ishikawa,Hironori Katoh,Manabu Negishi
The Journal of Neuroscience null
Proteomics strategy based on liquid-phase IEF and 2-D DIGE: application to bone marrow mesenchymal progenitor cells.
Beerelli Seshi
Proteomics null
Striatal enriched phosphatase 61 dephosphorylates Fyn at phosphotyrosine 420.
Tri-Hung Nguyen,Jian Liu,Paul J Lombroso
The Journal of Biological Chemistry null
Spatial learning enhances the expression of inositol 1,4,5-trisphosphate 3-kinase A in the hippocampal formation of rat.
Il Hwan Kim,Soon Kwon Park,Woong Sun,Yunhee Kang,Hyun Taek Kim,Hyun Kim
Brain Research Molecular Brain Research null
Gordana Miletic et al.
Pain, 149(1), 152-159 (2010-02-23)
Plasticity in the spinal dorsal horn may contribute to the development of pain following peripheral nerve injury. Shank proteins are a constituent family of the post-synaptic density (PSD), and they may play a role in synaptic plasticity through activity-dependent synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service