Skip to Content
Merck
All Photos(1)

Key Documents

L5530

Sigma-Aldrich

Latex beads, carboxylate-modified polystyrene, fluorescent orange

aqueous suspension, 0.5 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

form

aqueous suspension

Quality Level

composition

Solids, 2.5%

mean particle size

0.5 μm

fluorescence

λex ~520 nm; λem ~540 nm

Looking for similar products? Visit Product Comparison Guide

Application

Carboxylate-modified polystyrene latex beads (fluorescent orange) have been used to block phagocytosis by hemocytes in Drosophila. It has also been used to assess the uptake of latex beads by antigen presenting cells from porcine respiratory system.
Carboxylate-modified polystyrene latex beads have been used to collect morphofunctional data about the immune systems of Carabus lefebvrei, a helicophagous Italian endemic ground beetle often used as an indicator of the habitat quality of the Apennines mountain forests. Carboxylate-modified polystyrene latex beads have also been used to inform comparative studies on immunity defense mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa and to investigate the phagocytic activity of cultured rat thyroid cells.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Bajgar et al.
PLoS pathogens, 14(4), e1007022-e1007022 (2018-04-28)
Phagocytosis by hemocytes, Drosophila macrophages, is essential for resistance to Streptococcus pneumoniae in adult flies. Activated macrophages require an increased supply of energy and we show here that a systemic metabolic switch, involving the release of glucose from glycogen, is
Bente Winkler et al.
Science advances, 7(44), eabh0050-eabh0050 (2021-10-28)
[Figure: see text].
Yu Long Han et al.
Nature physics, 16(1), 101-108 (2020-09-10)
Sculpting of structure and function of three-dimensional multicellular tissues depend critically on the spatial and temporal coordination of cellular physical properties, yet the organizational principles that govern these events, and their disruption in disease, remain poorly understood. Using a multicellular
P G Giulianini et al.
Tissue & cell, 35(4), 243-251 (2003-08-19)
In the context of comparative studies on immunity defence mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa (Drury, 1770) the ultrastructure of the circulating hemocytes of the third instar larval stage has been investigated by means of light
Circulating hemocytes from larvae and adults of Carabus (Chaetocarabus) lefebvrei Dejean 1826 (Coleoptera, Carabidae): cell types and their role in phagocytosis after in vivo artificial non-self-challenge
Giglio, A., et al.
Micron (Oxford, England : 1993), 39, 52-58 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service