Merck
  • Brain inflammation triggers macrophage invasion across the blood-brain barrier in Drosophila during pupal stages.
Brain inflammation triggers macrophage invasion across the blood-brain barrier in Drosophila during pupal stages.
Science advances (2021-10-28)
Bente Winkler, Dominik Funke, Billel Benmimoun, Pauline Spéder, Simone Rey, Mary A Logan, Christian Klämbt
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green, aqueous suspension, 1.0 μm mean particle size
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent orange, aqueous suspension, 0.5 μm mean particle size