Skip to Content
Merck
All Photos(1)

Key Documents

L1764

Sigma-Aldrich

(±)-Lorazepam

Synonym(s):

7-Chloro-5-(2-chlorophenyl)-1,3-dihydro-3-hydroxy-2H-1,4-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10Cl2N2O2
CAS Number:
Molecular Weight:
321.16
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

originator

Baxter

SMILES string

OC1N=C(c2ccccc2Cl)c3cc(Cl)ccc3NC1=O

InChI

1S/C15H10Cl2N2O2/c16-8-5-6-12-10(7-8)13(19-15(21)14(20)18-12)9-3-1-2-4-11(9)17/h1-7,15,21H,(H,18,20)

InChI key

DIWRORZWFLOCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anxiolytic; ligand for the GABAA receptor benzodiazepine modulatory site.

Features and Benefits

This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Korkmaz et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 8(3), 175-181 (1998-08-26)
Rats were treated with lorazepam for four days (six times 2 mg kg(-1)) at the ages of 71, 118, 163 and 210 days. Increased excitation (physical dependence) in the withdrawal period was tested several times during 14 days with hexobarbital
Jason T McMullan et al.
Prehospital emergency care : official journal of the National Association of EMS Physicians and the National Association of State EMS Directors, 17(1), 1-7 (2012-11-15)
The choice of the optimal benzodiazepine to treat prehospital status epilepticus is unclear. Lorazepam is preferred in the emergency department, but concerns about nonrefrigerated storage limits emergency medical services (EMS) use. Midazolam is increasingly popular, but its heat stability is
Excess medicaid payments and the stock prices of drug companies.
William C Grant
Journal of managed care pharmacy : JMCP, 18(8), 650-650 (2012-11-07)
Olugbenga A Akingbola et al.
American journal of critical care : an official publication, American Association of Critical-Care Nurses, 21(6), 456-459 (2012-11-03)
A 6-year-old girl displayed symptoms of serotonin syndrome after accidental ingestion of Vyvanse (lisdexamfetamine dimesylate). Dexmedetomidine was administered because of persistent neuromuscular hyperactivity and severe agitation despite initial therapy with benzodiazepines. Some children show a paradoxical reaction to benzodiazepines, and
A 59-year-old woman who is awake yet unresponsive and stuporous after liver transplantation.
Insung Chung et al.
Chest, 143(4), 1163-1165 (2013-04-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service