KCQS06
KiCqStart® Probe qPCR ReadyMix™, ROX™
with ROX for ABI instruments
Synonym(s):
Kicqstart® Probe qPCR ReadyMix™
About This Item
Recommended Products
form
liquid
usage
sufficient for 1250 reactions
sufficient for 250 reactions
sufficient for 5000 reactions
feature
dNTPs included
hotstart
storage condition
protect from light
technique(s)
qPCR: suitable
color
light blue
input
purified DNA
compatibility
ABI 5700
ABI 7000
ABI 7300
ABI 7700
ABI 7900 HT Fast
ABI 7900 HT
ABI 7900
ABI StepOne
ABI StepOnePlus
detection method
probe-based
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
A key component of KiCqStart Probe qPCR ReadyMix is an ultra-pure, processive thermostable DNA polymerase that is free of detectable E. coli DNA§. This PCR master mix is ideal for demanding qPCR applications such as bacterial pathogen detection where residual host DNA in typical recombinant enzyme preparations can limit assay sensitivity and obscure detection of low copy samples. The enzyme in KiCqStart Probe qPCR ReadyMix is combined with high avidity monoclonal antibodies to provide a stringent automatic hot-start that allows reaction assembly, and temporary storage, at room temperature prior to PCR amplification.
Application
Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument.KiCqStart One-Step Probe RT-qPCR ReadyMix does not contain an internal reference dye.
Features and Benefits
- Assay results in as little as 33 minutes
- Highly efficient and sensitive real-time PCR results
- Little/no optimization required
Components
Legal Information
recommended
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.
Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.
Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.
Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.
Protocols
KiCqStart™ Probe qPCR ReadyMix™ is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments.
KiCqStart™ Probe qPCR ReadyMix™ is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments.
KiCqStart™ Probe qPCR ReadyMix™ is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments.
KiCqStart™ Probe qPCR ReadyMix™ is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service