Skip to Content
Merck
All Photos(2)

Key Documents

S7899

Sigma-Aldrich

Sodium acetate buffer solution

pH 5.2±0.1 (25 °C), for molecular biology, 3 M, 0.2 μm filtered

Synonym(s):

Sodium acetate buffer 3M

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3786729
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

liquid

concentration

3 M

pH

5.2±0.1 (25 °C)

suitability

suitable for DNA precipitation

storage temp.

room temp

SMILES string

[Na+].CC(O)=O.CC([O-])=O

InChI

1S/2C2H4O2.Na/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+1/p-1

InChI key

BHZOKUMUHVTPBX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sigma′s Sodium Acetate Buffer is a 3M solution prepared with 18 megaohm water and filter-sterilized.

Application

Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomas Erban et al.
Scientific reports, 7(1), 5084-5084 (2017-07-13)
Honeybee (Apis mellifera L.) workers act as passive vectors of Paenibacillus larvae spores, which cause the quarantine disease American foulbrood (AFB). We assessed the relative proportions of P. larvae within the honeybee microbiome using metabarcoding analysis of the 16 S rRNA
Sena Bluemel et al.
Gut microbes, 11(3), 265-275 (2019-04-16)
Alcohol-induced liver disease is closely related to translocation of bacterial products and bacteria from the intestine to the liver. However, it is not known whether bacterial translocation to the liver depends on certain intestinal microbiota changes that would predispose bacteria
Sambrook, J., and Russell, D.W.
Molecular Cloning: A Laboratory Manual, A1-A1 (2001)
M Strupp
Der Ophthalmologe : Zeitschrift der Deutschen Ophthalmologischen Gesellschaft, 110(1), 7-15 (2013-01-05)
The spectrum of diagnoses of patients with dizziness as the leading symptom who consult a neurologist does not differ greatly from the spectrum of those who consult ear nose and throat (ENT) specialists or general practitioners (GP). The most frequent
Tong Ihn Lee et al.
Nature protocols, 1(2), 729-748 (2007-04-05)
Genome-wide location analysis, also known as ChIP-Chip, combines chromatin immunoprecipitation and DNA microarray analysis to identify protein-DNA interactions that occur in living cells. Protein-DNA interactions are captured in vivo by chemical crosslinking. Cell lysis, DNA fragmentation and immunoaffinity purification of

Articles

Sodium Acetate Buffer provider. 5+ decades providing high-quality buffer products that maintain consistent performance for molecular biology .

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service