Skip to Content
Merck
All Photos(2)

Key Documents

HT1103128

Sigma-Aldrich

Eosin Y Solution, Alcoholic, with Phloxine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malgorzata Domino et al.
Theriogenology, 147, 18-24 (2020-02-20)
Equine endometrosis is a multifactorial chronic degenerative condition, considered to be one of a major causes of equine infertility. The formation of periglandular fibrosis seems to be linked to chronic inflammation of the mare endometrium in a paracrine way and
Jordi Miró et al.
Frontiers in veterinary science, 7, 631-631 (2020-11-03)
Collagen fibers and inflammatory cells are the basis for jenny endometrium Kenney and Doig's classification developed for the mare. The infiltration of a large number of eosinophils in the jenny endometrium is intriguing. Eosinophil and fibroblast produced IL33, which has
Maria Rosa Rebordão et al.
Theriogenology, 113, 8-18 (2018-02-18)
Neutrophil extracellular traps (NETs) are DNA complexes carrying nuclear and cytoplasmic proteins, such as elastase (ELA), cathepsin-G (CAT) and myeloperoxidase (MPO). Mare endometrosis is a chronic degenerative process characterized by excessive collagen in endometrium. While NETs fight bacteria that cause
Maria Rosa Rebordão et al.
Domestic animal endocrinology, 67, 1-10 (2018-12-07)
Prostaglandin E2 (PGE2) has contradictory effects in many organs. It may have proinflammatory, anti-inflammatory, or anti-fibrotic roles, depending on the type of receptors to which it binds. By signaling through its receptors EP2 and EP4, PGE2 mediates anti-inflammatory and anti-fibrotic
Brooke Tata et al.
Nature medicine, 24(6), 834-846 (2018-05-16)
Polycystic ovary syndrome (PCOS) is the main cause of female infertility worldwide and corresponds with a high degree of comorbidities and economic burden. How PCOS is passed on from one generation to the next is not clear, but it may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service