Skip to Content
Merck
All Photos(8)

Key Documents

HPA019081

Sigma-Aldrich

Anti-PDP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PDP 1, Anti-PDPC 1, Anti-PPM2C antibody produced in rabbit, Anti-Protein phosphatase 2C, Anti-Pyruvate dehydrogenase phosphatase, catalytic subunit 1, Anti-[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YIAVSLLPHETLLEIENAVESGRALLPILQWHKHPNDYFSKEASKLYFNSLRTYWQELIDLNTGESTDIDVKEALINAFKRLDNDISLEAQVGDPNSFLNYLVLRVAFSGATACVAHVD

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPM2C(54704)

General description

The gene pyruvate dehydrogenase phosphatase catalytic subunit-1 (PDP1) is mapped to human chromosome 8q22.1. It belongs to protein phosphatase 2C (PP2C) family. PDP1 is highly expressed in skeletal muscles.

Immunogen

[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pyruvate dehydrogenase complex (PDC) is important for glucose homeostasis. The core of PDC consists of pyruvate dehydrogenase phosphatase catalytic subunit-1 (PDP1) and pyruvate dehydrogenase E1 component subunit-α (PDHA1). PDP1 is down-regulated in patients with congenital lactic academia. PDP1 is also down-regulated in circulating lymphocytes of obese patients, signaling insulin resistance.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73650

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Fan et al.
Molecular cell, 53(4), 534-548 (2014-02-04)
Mitochondrial pyruvate dehydrogenase complex (PDC) is crucial for glucose homeostasis in mammalian cells. The current understanding of PDC regulation involves inhibitory serine phosphorylation of pyruvate dehydrogenase (PDH) by PDH kinase (PDK), whereas dephosphorylation of PDH by PDH phosphatase (PDP) activates
Mark van Duin et al.
Genes, chromosomes & cancer, 44(4), 438-449 (2005-09-01)
Copy number increase of 8q has previously been shown to be associated with a poor clinical outcome and tumor recurrence in patients with prostate cancer. In this study, a detailed genomic analysis of 8q was performed of archival primary and
Marco Piccinini et al.
Obesity research, 13(4), 678-686 (2005-05-18)
The objective of this study was to determine whether down-regulation of pyruvate dehydrogenase phosphatase (PDP) is responsible for poorly active pyruvate dehydrogenase (PDH) in circulating lymphocytes (CLs) of obese subjects (ObS), and if so, whether it improves when their plasma
Mary C Maj et al.
The Journal of clinical endocrinology and metabolism, 90(7), 4101-4107 (2005-04-28)
Pyruvate dehydrogenase phosphatase (PDP) deficiency has been previously reported as an enzymopathy, but the genetic basis for such a defect has never been established. The aim of this study was to identify the cause of the defect in two patients
M Ito et al.
Clinica chimica acta; international journal of clinical chemistry, 209(1-2), 1-7 (1992-07-31)
We developed an assay method for pyruvate dehydrogenase phosphatase activity using [1-14C]pyruvate and measured pyruvate dehydrogenase phosphatase activity in cultured skin fibroblasts from three patients with congenital lactic acidemia due to a defect in activation of the pyruvate dehydrogenase complex.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service