Skip to Content
Merck
All Photos(6)

Key Documents

HPA017887

Sigma-Aldrich

Anti-SLC36A4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Proton-coupled amino acid transporter 4, Anti-Proton/amino acid transporter 4, Anti-Solute carrier family 36 member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

REELDMDVMRPLINEQNFDGTSDEEHEQELLPVQKHYQLDDQEGISFVQTLMHLLKGNIGTGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 36 member 4 (SLC36A4) is a sodium-independent amino acid transporter which is present in lysosomes, endosomes and the plasma membrane. The gene encoding SLC36A4 is localized on chromosome 11.

Immunogen

Proton-coupled amino acid transporter 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC36A4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Solute carrier family 36 member 4 (SLC36A4) is a unique transporter in the cell which regulates growth and signaling pathways. On lysosomes and late endosomes, mechanistic target of rapamycin complex 1 (mTORC1) is activated by SLC36A4. In this, SLC36A4 targets the translation initiation factor 4E-binding protein 1 (4E-BP1). It has been shown that SLC36A4 stimulates the proliferation of colorectal cancer cells in vitro. Tryptophan is one of the substrates transported by SLC36A4.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73245

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aparna Kumar et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(51), 18249-18254 (2014-12-10)
Molecular biomarkers are changes measured in biological samples that reflect disease states. Such markers can help clinicians identify types of cancer or stages of progression, and they can guide in tailoring specific therapies. Many efforts to identify biomarkers consider genes
Samyuktha Muralidharan Pillai et al.
The Journal of biological chemistry, 286(4), 2455-2460 (2010-11-26)
The SLC36 family of transporters consists of four genes, two of which, SLC36A1 and SLC36A2, have been demonstrated to code for human proton-coupled amino acid transporters or hPATs. Here we report the characterization of the fourth member of the family
S Heublein et al.
Oncogene, 29(28), 4068-4079 (2010-05-26)
The phosphoinositide3-kinase (PI3K)/Akt and downstream mammalian target of rapamycin complex 1 (mTORC1) signalling cascades promote normal growth and are frequently hyperactivated in tumour cells. mTORC1 is also regulated by local nutrients, particularly amino acids, but the mechanisms involved are poorly
S-J Fan et al.
Oncogene, 35(23), 3004-3015 (2015-10-06)
Tumour cells can use strategies that make them resistant to nutrient deprivation to outcompete their neighbours. A key integrator of the cell's responses to starvation and other stresses is amino-acid-dependent mechanistic target of rapamycin complex 1 (mTORC1). Activation of mTORC1
Yuying Liu et al.
Cancer cell, 33(3), 480-494 (2018-03-14)
Despite the clinical successes fostered by immune checkpoint inhibitors, mechanisms underlying PD-1 upregulation in tumor-infiltrating T cells remain an enigma. Here, we show that tumor-repopulating cells (TRCs) drive PD-1 upregulation in CD8+ T cells through a transcellular kynurenine (Kyn)-aryl hydrocarbon receptor (AhR)

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service