Merck
  • Tumor-Repopulating Cells Induce PD-1 Expression in CD8+ T Cells by Transferring Kynurenine and AhR Activation.
Tumor-Repopulating Cells Induce PD-1 Expression in CD8+ T Cells by Transferring Kynurenine and AhR Activation.
Cancer cell (2018-03-14)
Yuying Liu, Xiaoyu Liang, Wenqian Dong, Yi Fang, Jiadi Lv, Tianzhen Zhang, Roland Fiskesund, Jing Xie, Jinyan Liu, Xiaonan Yin, Xun Jin, Degao Chen, Ke Tang, Jingwei Ma, Huafeng Zhang, Jing Yu, Jun Yan, Huaping Liang, Siqi Mo, Feiran Cheng, Yabo Zhou, Haizeng Zhang, Jing Wang, Jingnan Li, Yang Chen, Bing Cui, Zhuo-Wei Hu, Xuetao Cao, F Xiao-Feng Qin, Bo Huang
ABSTRACT
Despite the clinical successes fostered by immune checkpoint inhibitors, mechanisms underlying PD-1 upregulation in tumor-infiltrating T cells remain an enigma. Here, we show that tumor-repopulating cells (TRCs) drive PD-1 upregulation in CD8+ T cells through a transcellular kynurenine (Kyn)-aryl hydrocarbon receptor (AhR) pathway. Interferon-γ produced by CD8+ T cells stimulates release of high levels of Kyn produced by TRCs, which is transferred into adjacent CD8+ T cells via the transporters SLC7A8 and PAT4. Kyn induces and activates AhR and thereby upregulates PD-1 expression. This Kyn-AhR pathway is confirmed in both tumor-bearing mice and cancer patients and its blockade enhances antitumor adoptive T cell therapy efficacy. Thus, we uncovered a mechanism of PD-1 upregulation with potential tumor immunotherapeutic applications.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-IDO (Indoleamine 2,3-Dioxygenase) Antibody, clone 10.1, clone 10.1, Upstate®, from mouse
Sigma-Aldrich
Rabbit anti-TopBP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken
Sigma-Aldrich
Indoxyl sulfate potassium salt
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
Anti-SLC36A4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution