Skip to Content
Merck
All Photos(7)

Key Documents

HPA014791

Sigma-Aldrich

Anti-CLPTM1L antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CLPTM1-like protein, Anti-CRR9p, Anti-Cisplatin resistance-related protein 9, Anti-Cleft lip and palate transmembrane protein 1-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LNVEDFDVESKFERTVNVSVPKKTRNNGTLYAYIFLHHAGVLPWHDGKQVHLVSPLTTYMVPKPEEINLLTGESDTQQIEAEKKPTSALDEPVSHWRPRLALNVMADNFVFDGSSLPADVHRYMKMIQLGKTVHYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLPTM1L(81037)

General description

CLPTM1L (cleft lip and palate trans-membrane 1-like) was originally discovered as a gene conferring resistance to cisplatin in ovarian cancer cells. Thus, it is also called cisplatin resistance-related protein 9 (CRR9p). This gene is localized to human chromosome 5p15.33. It is expressed in multiple types of tumors, and is up-regulated in cisplatin-resistant tumor cell lines.

Immunogen

Cleft lip and palate transmembrane protein 1-like protein recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

CLPTM1L (cleft lip and palate trans-membrane 1-like) is mainly involved in maintaining genomic stability and integrity, by controlling the activity of telomerases. This protein might be involved in apoptosis. It is up-regulated in lung cancer, and regulates Bcl-xL, and inhibits genotoxic stress-mediated apoptosis in lung tumor cells. Therefore, it facilitates lung tumorigenesis. It facilitates growth in pancreatic cells, and is up-regulated in pancreatic cancer cells, where it promotes growth and aneuploidy. Genome wide association studies (GWAS) show that polymorphisms in this gene are also linked to susceptibility to familial and bilateral testicular germ cell tumours (TGCTs).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73014

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojie Xun et al.
Medicine, 93(28), e289-e289 (2014-12-20)
Genetic variants of cleft lip and palate trans-membrane 1-like (CLPTM1L) genes in the p15.33 region of chromosome 5 were previously identified to influence susceptibility to lung cancer. We examined the association of single nucleotide polymorphisms (SNPs) in CLPTM1L genes with
Stephanie Trezise et al.
International journal of molecular sciences, 19(8) (2018-07-26)
Antibody Secreting Cells (ASCs) are a fundamental component of humoral immunity, however, deregulated or excessive antibody production contributes to the pathology of autoimmune diseases, while transformation of ASCs results in the malignancy Multiple Myeloma (MM). Despite substantial recent improvements in
Yunwen Hou et al.
Cell transplantation, 30, 9636897211045970-9636897211045970 (2021-09-30)
This study aimed to explore the function of CLPTM1L in oral squamous cell carcinoma and mechanism of tumorigenesis. The expression of CLPTM1L was detected by immunohistochemistry. The localization in cells was detected by immunofluorescence. Cell invasion, proliferation, and migration were
Hang Li et al.
Cell communication and signaling : CCS, 18(1), 152-152 (2020-09-19)
Radioresistance is a major challenge in lung cancer radiotherapy, and new radiosensitizers are urgently needed. Estrogen receptor β (ERβ) is involved in the progression of non-small cell lung cancer (NSCLC), however, the role of ERβ in the response to radiotherapy
László G Puskás et al.
Molecular cancer therapeutics, 15(5), 985-997 (2016-03-05)
We and others have recently shown cisplatin resistance-related protein 9 (CRR9)/Cleft Lip and Palate Transmembrane 1-Like (CLPTM1L) to affect survival and proliferation in lung and pancreatic tumor cells. Our research has indicated that CLPTM1L affects multiple survival signaling pathways in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service