Skip to Content
Merck
All Photos(3)

Key Documents

HPA010636

Sigma-Aldrich

Anti-CD164 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD164 antigen antibody produced in rabbit, Anti-MGC-24 antibody produced in rabbit, Anti-MUC-24 antibody produced in rabbit, Anti-Multi-glycosylated core protein 24 antibody produced in rabbit, Anti-Putative mucin core protein 24 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20-1:50
western blot: 0.04-0.4 μg/mL

immunogen sequence

TQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATPVPTANSTAKPTVQPSPSTTSKTVTTSGTTNNTVTPTS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Gene Information

human ... CD164(8763)

Immunogen

Putative mucin core protein 24 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Sialomucin core protein 24 (also known as endolyn or CD164) is a protein encoded by the CD164 gene in humans. It is a sialomucin that plays roles in regulating proliferation, adhesion, and differentiation of hematopoietic stem cells. CD164 may act as a new target for diagnosis and treatment for colon cancer. It participates in regulating the proliferation, cell adhesion and differentiation of hematopoietic stem and progenitor cells. It is also involved in the development of cancer. The protein functions as a receptor that regulates stem cell localization to the bone marrow and may participate in the localization of prostate cancer cells to the marrow.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71974

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin-Wu Zhang et al.
Oncology reports, 44(6), 2419-2428 (2020-10-31)
Colon cancer (CC) is one of the leading causes of cancer‑related mortality in China and western countries. Several studies have demonstrated that long non‑coding RNAs (lncRNAs) play critical roles in cancer development. However, the function of lncRNA RP11‑619L19.2 in colon
A M Havens et al.
BMC cancer, 6, 195-195 (2006-07-25)
The chemokine stromal derived factor-1 (SDF-1 or CXCL12) and its receptor CXCR4 have been demonstrated to be crucial for the homing of stem cells and prostate cancers to the marrow. While screening prostate cancers for CXCL12-responsive adhesion molecules, we identified
Jingqun Tang et al.
BMC molecular biology, 12, 44-44 (2011-10-18)
CD164 (also known as MGC-24v or endolyn) is a sialomucin which has been suggested to participate in regulating the proliferation, cell adhesion and differentiation of hematopoietic stem and progenitor cells. CD164 is also involved in the development of cancer. The
Jia-Hong Chen et al.
Oncology letters, 14(5), 6103-6108 (2017-11-09)
The aim of the present study was to investigate the association between cluster of differentiation (CD) 164 expression with clinicopathological parameters and prognosis among patients with oral cavity squamous cell carcinoma (OSCC). The present study retrospectively reviewed 70 patients with
Jingqun Tang et al.
Cancer investigation, 30(5), 380-389 (2012-03-14)
CD164 (Endolyn) is a sialomucin, which has been found to play roles in regulating proliferation, adhesion, and differentiation of hematopoietic stem cells. Possible association of CD164 with solid cancer development remains unknown. We first studied CD164 expression in biopsies from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service