Skip to Content
Merck
All Photos(6)

Key Documents

HPA006889

Sigma-Aldrich

Anti-SORT1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-100 kDa NT receptor antibody produced in rabbit, Anti-Glycoprotein 95 antibody produced in rabbit, Anti-Gp95 antibody produced in rabbit, Anti-NT3 antibody produced in rabbit, Anti-NTR3 antibody produced in rabbit, Anti-Neurotensin receptor 3 antibody produced in rabbit, Anti-Sortilin precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LERNCEEKDYTIWLAHSTDPEDYEDGCILGYKEQFLRLRKSSVCQNGRDYVVTKQPSICLCSLEDFLCDFGYYRPENDSKCVEQPELKGHDLEFCLYGREEHLTTNGYRKIPGDKCQGGVNPVREVKD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORT1(6272)

Immunogen

Sortilin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SORT1 (Sortilin 1) is a sorting receptor protein. It encodes sortilin that participates in the metabolism of low-density lipoprotein cholesterol (LDL-C) in plasma. It helps to relocate various target proteins including growth factors, signaling receptors, and enzymes in the specific secretory or endocytic cell compartment. It maintains the activities of neuronal and non-neuronal cell types. Malfunctioning of this gene is responsible for cardiovascular and neurological diseases. It even can cause malignancies, including hypercholesterolemia, retinal degeneration, neuronal cell loss, Alzheimer′s disease. It also has potentiality to mediate the endocytosis and degradation of lipoprotein lipase expressed on the cell surface.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70149

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Sophie Carlo et al.
Journal of molecular medicine (Berlin, Germany), 92(9), 905-911 (2014-05-20)
Sortilin is a sorting receptor that directs target proteins, such as growth factors, signaling receptors, and enzymes, to their destined location in secretory or endocytic compartments of cells. The activity of sortilin is essential for proper function of not only
Genevieve E Campbell et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(4), e21481-e21481 (2021-03-13)
The midcycle luteinizing hormone (LH) surge initiates a cascade of events within the ovarian follicle which culminates in ovulation. Only mural granulosa cells and theca cells express large numbers of LH receptors, and LH-stimulated paracrine mediators communicate the ovulatory signal
Maria Francisca Coutinho et al.
Revista portuguesa de cardiologia : orgao oficial da Sociedade Portuguesa de Cardiologia = Portuguese journal of cardiology : an official journal of the Portuguese Society of Cardiology, 32(10), 793-799 (2013-08-06)
Plasma low-density lipoprotein cholesterol (LDL-C) levels are a key determinant of the risk of cardiovascular disease, which is why many studies have attempted to elucidate the pathways that regulate its metabolism. Novel latest-generation sequencing techniques have identified a strong association
M S Nielsen et al.
The Journal of biological chemistry, 274(13), 8832-8836 (1999-03-20)
Lipoprotein lipase and the receptor-associated protein (RAP) bind to overlapping sites on the low density lipoprotein receptor-related protein/alpha2-macroglobulin receptor (LRP). We have investigated if lipoprotein lipase interacts with the RAP binding but structurally distinct receptor sortilin/neurotensin receptor-3. We show, by
Chun-Chi Chen et al.
Oncotarget, 8(44), 77152-77167 (2017-11-05)
Loss of immunosurveillance is a major cause of cancer progression. Here, we demonstrate that gelsolin, a constituent of ejaculate, induces apoptosis of activated lymphocytes in prostate cancer. Gelsolin was highly expressed in prostate cancer cells, and was associated with tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service