Skip to Content
Merck
All Photos(5)

Key Documents

HPA003827

Sigma-Aldrich

Anti-F13B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Coagulation factor XIII B chain precursor antibody produced in rabbit, Anti-Fibrin-stabilizing factor B subunit antibody produced in rabbit, Anti-Protein-glutamine γ-glutamyltransferase B chain antibody produced in rabbit, Anti-Transglutaminase B chain antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FCLAGYTTESGRQEEQTTCTTEGWSPEPRCFKKCTKPDLSNGYISDVKLLYKIQENMHYGCASGYKTTGGKDEEVVQCLSDGWSSQPTCRKEHETCLAPELYNGNYSTTQKTFKVKDKVQYECATGYYTAGGKKTEEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... F13B(2165)

Immunogen

Coagulation factor XIII B chain precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Factor XIII (FXIII) is a heterotetramer, fibrin-stabilising factor consisting of catalytic A subunits (FXIII-A or F13A) and carrier B subunits (FXIII-B or F13B). FXIII-B consists of 10 sushi domains. F13B helps in preventing the fast clearance of F13A from the circulation. Both genes cause inherited severe FXIII deficiency, which is a rare coagulation disorder. Factor XIII helps in stabilizing fibrin and protecting it from fibrinolysis, it is also involved in wound healing and angiogenesis. F13B plays a carrier role for zymogen F13A, which is highly polymorphic.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84498

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideho Wada et al.
Thrombosis and haemostasis, 109(4), 661-668 (2013-02-15)
Factor XIII (FXIII) is a fibrin-stabilising factor consisting of catalytic A subunits (FXIII-A) and carrier B subunits (FXIII-B). FXIII-B prevents the fast clearance of FXIII-A from the circulation. Congenital FXIII-A deficiency is a rare bleeding disorder, and congenital FXIII-B deficiency
A Biswas et al.
Hamostaseologie, 34(2), 160-166 (2014-02-08)
The plasma circulating zymogenic coagulation factor XIII (FXIII) is a protransglutaminase, which upon activation by thrombin and calcium cross-links preformed fibrin clots/fibrinolytic inhibitors making them mechanically stable and less susceptible to fibrinolysis. The zymogenic plasma FXIII molecule is a heterotetramer
James R Byrnes et al.
Blood, 128(15), 1969-1978 (2016-08-27)
Coagulation transglutaminase factor XIII (FXIII) exists in circulation as heterotetrameric proenzyme FXIII-A
Masayoshi Souri et al.
Biochemistry, 47(33), 8656-8664 (2008-07-26)
Factor XIII (FXIII) is a heterotetramer composed of two catalytic A subunits (FXIII-A) and two B subunits (FXIII-B). FXIII-B has 10 Sushi domains. To explore the structure-function relationship of FXIII-B, we looked for domains in FXIII-B responsible for its homodimer
N Komanasin et al.
Journal of thrombosis and haemostasis : JTH, 3(11), 2487-2496 (2005-10-26)
Factor (F)XIII B-subunit, which plays a carrier role for zymogen FXIIIA, is highly polymorphic, but the molecular basis for these polymorphisms and their relationship to disease remains unknown. To screen the FXIIIB gene coding region for common variation and analyze

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service