Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200048

Sigma-Aldrich

Anti-E1B-AP5 antibody, Rat monoclonal

clone EAP4A11, purified from hybridoma cell culture

Synonym(s):

Anti-Adenovirus E1B 55-KD protein-associated protein 5, Anti-Heterogeneous nuclear ribonucleoprotein U-like 1, Anti-hnRNPUL1, Anti-hnRPUL1, Monoclonal Anti-E1B-AP5 antibody produced in rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

EAP4A11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

bovine, canine, human, rat, monkey, mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.05-0.1 μg/mL using HT-29 cell extracts

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-E1B-AP5 (rat IgG2b isotype) is derived from the hybridoma EAP4A11 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from rat immunized with full length human E1B-AP5 fusion protein. E1B-55K-associated protein 5 (E1B-AP5), also known as heterogeneous nuclear ribonucleoprotein U-like 1(hnRNPUL1), is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family.
The E1B-AP5 gene is located on the human chromosome at 19q13.2.

Specificity

Monoclonal Anti-E1B-AP5 recognizes human, monkey, bovine, dog, mouse and rat E1B-AP5.

Immunogen

full length human E1B-AP5 fusion protein.

Application

Monoclonal Anti-E1B-AP5 antibody produced in rat may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

E1B-55K-associated protein 5 (E1B-AP5) plays a vital role in ATR signaling. In addition, it may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B55kDa in adenovirus-infected cells. Interestingly, this protein was found to be associated with bromodomain-containing protein, BRD7 in a complex that links chromatin events with mRNA processing at the level of transcriptional regulation. It also activates transcription of glucocorticoid-responsive promoter in the absence of ligand-stimulation.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2-8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interaction of the hnRNP family member E1B-AP5 with p53
Barral PM, et al.
Febs Letters, 579, 2752-2758 (2005)
Kentaro Ohki et al.
Haematologica, 104(1), 128-137 (2018-09-02)
Fusion genes involving MEF2D have recently been identified in precursor B-cell acute lymphoblastic leukemia, mutually exclusive of the common risk stratifying genetic abnormalities, although their true incidence and associated clinical characteristics remain unknown. We identified 16 cases of acute lymphoblastic
Regulation of transcription by the heterogeneous nuclear ribonucleoprotein E1B-AP5 is mediated by complex formation with the novel bromodomain-containing protein BRD7.
Kzhyshkowska J, et al.
The Biochemical Journal, 371, 385-385 (2003)
Regulation of DNA-End Resection by hnRNPU-like Proteins Promotes DNA Double-Strand Break Signaling and Repair
Polo SE, et al.
Molecular Cell, 45, 505-516 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service