Skip to Content
Merck
All Photos(6)

Key Documents

HPA003564

Sigma-Aldrich

Anti-FCGBP antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fcγ-binding protein antigen antibody produced in rabbit, Anti-FcγBP antibody produced in rabbit, Anti-IgGFc-binding protein precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LQAGDVVEFEVRPSWPLYLSANVGIQVLLFGTGAIRNEVTYDPYLVLIPDVAAYCPAYVVKSVPGCEGVALVVAQTKAISGLTIDGHAVGAKLTWEAVPGSEFSYAEVELGTADMIHTAEATTNLGLLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCGBP(8857)

Looking for similar products? Visit Product Comparison Guide

Immunogen

IgGFc-binding protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FCGBP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

IgGFc-binding protein is a protein encoded by the FCGBP gene in humans. The gene is also known as FcγBP. Its mRNA is expressed only in placenta and colonic epithelial cells. It may play an important role in immune protection and inflammation in the intestines of primates. The gene is expressed in both human and mouse prostates. Differential expression of this gene could reflect its potential role in prostate malignancy as well as an indicator for progression of the cancer. It is differentially expressed in normal thyroid tissue, thyroid adenomas and thyroid carcinomas. The gene is constitutively expressed in normal thyroid tissue; its expression is significantly increased in follicular thyroid adenomas and significantly decreased in papillary and follicular thyroid carcinomas. The expression level of this gene in thyroid biopsies is useful for distinguishing between a thyroid follicular adenoma and a follicular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86489

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziming Yuan et al.
OncoTargets and therapy, 14, 967-977 (2021-02-20)
The liver is the most frequent site for metastatic spread in colorectal cancer (CRC) patients, and these patients have much poorer prognosis than those without metastasis. Previous studies have shown that IgG Fc binding protein (FCGBP) plays important roles in
Qin Ding et al.
Scientific reports, 14(1), 4426-4426 (2024-02-24)
In head and neck squamous cell carcinoma (HNSC), chemoresistance is a major reason for poor prognosis. Nevertheless, there is a lack of validated biomarkers to screen for patients for categorical chemotherapy. Fc gamma binding protein (FCGBP) is a mucus protein
Timo K Albert et al.
Journal of proteome research, 9(6), 3108-3117 (2010-04-29)
TFF3 is a secretory peptide belonging to the trefoil factor family with a predicted size of 59 amino acid residues containing seven cysteine residues. It is predominantly expressed in intestinal goblet cells where it plays a key role in mucosal
N O'Donovan et al.
The Journal of endocrinology, 174(3), 517-524 (2002-09-05)
The genetic events involved in thyroid carcinogenesis are still incompletely understood. Several rearrangements and mutations of oncogenes have been implicated in the development of thyroid papillary carcinomas, follicular adenomas and carcinomas. However, none of these molecular alterations is suitable either
Mozammel H Gazi et al.
Cancer biology & therapy, 7(1), 70-75 (2007-10-17)
By means of protein expression profile, mass spectral and/or RT-PCR analyses we found for the first time IgG Fc binding protein (Fc gammaBP), distinct from Fc gamma receptors is expressed in both human and mouse prostates. There is a strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service