Skip to Content
Merck
All Photos(5)

Key Documents

HPA003415

Sigma-Aldrich

Anti-LTBP2 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

LTBP2 Antibody - Anti-LTBP2 antibody produced in rabbit, Ltbp2 Antibody, Anti-LTBP-2 antibody produced in rabbit, Anti-Latent-transforming growth factor β-binding protein 2 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

ERSPNLRRSSAAGEGTLARAQPPAPQSPPAPQSPPAGTLSGLSQTHPSQQHVGLSRTVRLHPTATASSQLSSNALPPGPGLEQRDGTQQAVPLEHPSSPWGLNLTEKIKKIKIVFTPTICKQTCARGHCANSCERG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LTBP2(4053)

Immunogen

Latent-transforming growth factor β-binding protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Latent-transforming growth factor β-binding protein 2 is a protein encoded by the LTBP2 gene in humans and is localized to chromosome 14q24. It is synthesized as a 240kDa protein by human foreskin fibroblasts. In response to tension stress, it may negatively control the function of fibulin-5, thereby modulating the mechanism of oxytalan fiber coalescence. Mutation in this gene causes primary congenital glaucoma (PCG). Other disorders manifest as secondary glaucoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70611

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eichi Tsuruga et al.
Connective tissue research, 53(6), 521-527 (2012-07-26)
Oxytalan fibers are extracellular matrix components consisting of pure microfibrils. However, the mechanism whereby oxytalan fibers develop is not fully understood. We have previously reported that in human periodontal ligament (PDL) fibroblasts subjected to stretching stress, bundles of oxytalan fibers
A Morén et al.
The Journal of biological chemistry, 269(51), 32469-32478 (1994-12-23)
Latent transforming growth factor-beta (TGF-beta)-binding protein (LTBP) is a component of the latent TGF-beta complex in human platelets. LTBP is composed of two different cysteine-rich repeat sequences, i.e. epidermal growth factor (EGF)-like repeats and a repeat containing 8 cysteine residues.
Sahar Jelodari-Mamaghani et al.
Molecular vision, 19, 333-347 (2013-02-13)
To assess for the first time the possible contribution of latent transforming growth factor (TGF)-beta binding protein 2 (LTBP2), an extracellular matrix (ECM) protein that associates with fibrillin-1-containing microfibrils, to the etiology of primary open angle glaucoma (POAG) and pseudoexfoliation
Zilong Li et al.
Oncogenesis, 8(11), 66-66 (2019-11-07)
Malignant lung cancer cells are characterized by uncontrolled proliferation and migration. Aberrant lung cancer cell proliferation and migration are programmed by altered cancer transcriptome. The underlying epigenetic mechanism is unclear. Here we report that expression levels of BRG1, a chromatin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service