Skip to Content
Merck
All Photos(2)

Key Documents

HPA001800

Sigma-Aldrich

Anti-UPF3B antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HUPF3B, Anti-MRX62, Anti-RENT3B, Anti-UPF3X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AKKLDKENLSDERASGQSCTLPKRSDSELKDEKPKRPEDESGRDYREREREYERDQERILRERERLKRQEEERRRQKERYEKEKTFKRKEEEMKKEKDTLRDKGKKAESTESIGSSEKTEKKEEVVKRDRIRNKDR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UPF3B(65109)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Regulator of nonsense transcripts 3B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

UPF3B (UPF3 regulator of nonsense transcripts homolog B) is a nucleo-cytoplasmic shuttling protein. It is expressed in all adult tissues. It shuttles between nucleus and cytoplasmic region. It consists of a conserved domain that participates in interaction with the exon junction complex (EJC) protein Y14. Furthermore, nonsense-mediated mRNA decay (NMD) mechanism is highly dependent on the Y14/UPF3B interaction. In Y14/UPF3B interaction pathway, EJC proteins act as adhering points for the formation of distinguishable NMD-activating mRNPs. For instance, during NMD UPF3B binds to the spliced β-globin mRNA in vivo to eliminate mRNAs containing a premature translation termination codon.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74305

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels H Gehring et al.
Molecular cell, 20(1), 65-75 (2005-10-08)
Messenger RNAs (mRNAs) bearing premature translation termination codons (PTCs) are degraded by nonsense-mediated mRNA decay (NMD). For mammalian NMD, current models propose a linear pathway that involves the splicing-dependent deposition of exon-junction complexes (EJCs) and the sequential action of the
Shihua Lu et al.
RNA biology, 1(1), 42-47 (2006-12-30)
The subcellular localization of the nonsense mediated decay (NMD) of mRNA transcripts bearing premature termination codons has been controversial. Recently, it has been demonstrated that RNA tethering of key mediators of NMD, including human UPF3B, accurately recreates NMD. Here, we
Malwina Michalak et al.
International journal of molecular sciences, 21(15) (2020-07-29)
DNA mismatch repair-deficient colorectal cancers (CRCs) accumulate numerous frameshift mutations at repetitive sequences recognized as microsatellite instability (MSI). When coding mononucleotide repeats (cMNRs) are affected, tumors accumulate frameshift mutations and premature termination codons (PTC) potentially leading to truncated proteins. Nonsense-mediated
Hala Chamieh et al.
Nature structural & molecular biology, 15(1), 85-93 (2007-12-11)
Nonsense-mediated mRNA decay (NMD) eliminates mRNAs containing a premature translation termination codon through the recruitment of the conserved NMD factors UPF1, UPF2 and UPF3. In humans, a dynamic assembly pathway allows UPF1 to join UPF2 and UPF3 recruited to the
J Lykke-Andersen et al.
Cell, 103(7), 1121-1131 (2001-02-13)
Nonsense-mediated decay (NMD) rids eukaryotic cells of aberrant mRNAs containing premature termination codons. These are discriminated from true termination codons by downstream cis-elements, such as exon-exon junctions. We describe three novel human proteins involved in NMD, hUpf2, hUpf3a, and hUpf3b.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service