Skip to Content
Merck
All Photos(5)

Key Documents

HPA001223

Sigma-Aldrich

Anti-CBS antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Beta- thionase, Anti-Cystathionine beta-synthase, Anti-Serine sulfhydrase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GGSVKDRISLRMIEDAERDGTLKPGDTIIEPTSGNTGIGLALAAAVRGYRCIIVMPEKMSSEKVDVLRALGAEIVRTPTNARFDSPESHVGVAWRLKNEIPNSHILDQYRNASNPLAHYDTTADEILQQCDGKLDMLVASVGTGGTITGIGAPHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBS(875)

General description

CBS (cystathionine-β-synthase) contains amino-terminal heme binding, central catalytic, and carboxyl-terminal regulatory domains.

Immunogen

Cystathionine beta-synthase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CBS (cystathionine-β-synthase) encodes a heme containing protein that catalyzes the conversion of serine and homocysteine to cystathionine, which is the first step in the transsulfuration pathway. It acts as a homotetramer and uses pyridoxal phosphate as a cofactor. S-adenosylmethionine (SAM) binding to CBS activates catalytic domain of CBS. Defects in this gene lead to deficiency of CBS causing homocystinuria, which is characterized by increased levels of the toxic metabolite homocysteine. Higher levels of homocysteine are associated with increased risk for cardiovascular diseases, Alzheimer′s disease and with neural tube defects. Polymorphism in it is also linked to bipolar disorder. Presence of CBS in human fallopian tube epithelium is crucial for production of gaseotransmitter, hydrogen sulphide, which relaxes the spontaneous contraction of the human oviduct. It is up-regulated in colon cancer cells and supports tumor growth and stimulates angiogenesis and vasorelaxation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82418

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nannan Ning et al.
Nature communications, 5, 4107-4107 (2014-06-11)
Embryo retention in the fallopian tube is thought to lead to ectopic pregnancy, which is a significant cause of morbidity. Hydrogen sulphide (H2S) is a gaseotransmitter produced mainly by cystathionine-γ-lyase and cystathionine-β-synthase. Here we show that cystathionine-γ-lyase and cystathionine -β-synthase
Thomas J McCorvie et al.
The Journal of biological chemistry, 289(52), 36018-36030 (2014-10-23)
Cystathionine β-synthase (CBS) is a key enzyme in sulfur metabolism, and its inherited deficiency causes homocystinuria. Mammalian CBS is modulated by the binding of S-adenosyl-l-methionine (AdoMet) to its regulatory domain, which activates its catalytic domain. To investigate the underlying mechanism
Agnieszka Permoda-Osip et al.
Neuropsychobiology, 69(2), 107-111 (2014-03-01)
Increased levels of homocysteine have been observed in various psychiatric disorders, among them in schizophrenia, depression and bipolar mood disorder. Of the genes connected with homocysteine metabolism, some studies have found an association between polymorphism of the methylenetetrahydrofolate reductase (MTHFR)
Katalin Módis et al.
Nitric oxide : biology and chemistry, 41, 146-156 (2014-03-29)
Recent data show that colon cancer cells selectively overexpress cystathionine-β-synthase (CBS), which produces hydrogen sulfide (H2S), to maintain cellular bioenergetics, support tumor growth and stimulate angiogenesis and vasorelaxation in the tumor microenvironment. The purpose of the current study was to
M Meier et al.
The EMBO journal, 20(15), 3910-3916 (2001-08-03)
Cystathionine beta-synthase (CBS) is a unique heme- containing enzyme that catalyzes a pyridoxal 5'-phosphate (PLP)-dependent condensation of serine and homocysteine to give cystathionine. Deficiency of CBS leads to homocystinuria, an inherited disease of sulfur metabolism characterized by increased levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service