Skip to Content
Merck
All Photos(4)

Key Documents

H9633

Sigma-Aldrich

L-Homocysteic acid

≥98% (TLC)

Synonym(s):

L-2-Amino-4-sulfobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO5S
CAS Number:
Molecular Weight:
183.18
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

Product Name

L-Homocysteic acid, ≥95%

Quality Level

Assay

≥95%
≥98% (TLC)

form

powder

color

white to off-white

application(s)

cell analysis

SMILES string

N[C@@H](CCS(O)(=O)=O)C(O)=O

InChI

1S/C4H9NO5S/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H,8,9,10)/t3-/m0/s1

InChI key

VBOQYPQEPHKASR-VKHMYHEASA-N

Application

L-Homocysteic acid has been used to study hydrogen peroxide-mediated changes in physiological and pathological conditions in the nervous system.

Biochem/physiol Actions

L-Homocysteic acid (L-HCA) is an endogenous neurotransmitter ligand of NMDA (N-methyl-D-aspartate) receptors. It is an analog of glutamate and competitively suppresses glutamate-cystine (xc-) antiporter function in neurons, causing decrease in cysteine uptake.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hari H Subramanian et al.
The Journal of comparative neurology, 526(2), 229-242 (2017-09-07)
The nucleus retroambiguus (NRA) is a neuronal cell group in the medullary ventrolateral tegmentum, rostrocaudally between the obex and the first cervical spinal segment. NRA neurons are premotor interneurons with direct projections to the motoneurons of soft palate, pharynx, and
M Piraud et al.
Rapid communications in mass spectrometry : RCM, 19(22), 3287-3297 (2005-10-20)
Seventy-six compounds of biological interest for the diagnosis of inherited disorders of amino acids (AA) metabolism have previously been demonstrated to be detectable in positive mode electrospray ionisation tandem mass spectrometry (ESI-MS/MS), after separation by ion-pairing reversed-phase liquid chromatography (RPLC).
J W Olney et al.
Brain research bulletin, 19(5), 597-602 (1987-11-01)
L-Homocysteic acid (L-HCA) has been proposed as a natural transmitter at the N-methyl-D-aspartate (NMDA) subtype of excitatory amino acid receptor based on recent evidence that L-HCA occurs L-HCA occurs naturally in the mammalian CNS, is released from K+ stimulated brain
T Knöpfel et al.
Neuroscience letters, 81(1-2), 188-192 (1987-10-16)
The effects of ionophoretically applied L-homocysteate (L-HCA), L-glutamate (L-Glu) and N-methyl-D-aspartate (NMDA) were compared in rat neocortical neurons recorded intracellularly in vitro. The firing pattern and the time course of membrane depolarization induced by L-HCA resembled those of NMDA responses.
Itopa E Ajayi et al.
Respiratory physiology & neurobiology, 252-253, 18-27 (2018-03-20)
Changes in cardiorespiratory control accompany the expression of complex emotions, indicative of limbic brain inputs onto bulbar autonomic pathways. Previous studies have focussed on the role of the prefrontal cortex in autonomic regulation. However, the role of the hippocampus, also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service