Skip to Content
Merck
All Photos(1)

Key Documents

H7375

Sigma-Aldrich

Histamine bisphosphate monohydrate

Synonym(s):

2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(4-Imidazolyl)ethylamine diphosphate salt monohydrate, Histamine diphosphate salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3 · 2H3PO4 · H2O
CAS Number:
Molecular Weight:
325.15
Beilstein:
3744619
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

storage temp.

−20°C

SMILES string

[H]O[H].OP(O)(O)=O.OP(O)(O)=O.NCCc1c[nH]cn1

InChI

1S/C5H9N3.2H3O4P.H2O/c6-2-1-5-3-7-4-8-5;2*1-5(2,3)4;/h3-4H,1-2,6H2,(H,7,8);2*(H3,1,2,3,4);1H2

InChI key

CNSGLKZGHLZDHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Shall et al.
Acta dermato-venereologica, 71(3), 199-204 (1991-01-01)
Terfenadine is a safe non-sedative H1-receptor antagonist. This study aimed to quantify the relative reduction in weal and flare area, thickness and erythema at 4, 8, 12 and 24 h following a single but variable oral dose of terfenadine compared
H S Nelson et al.
The Journal of allergy and clinical immunology, 92(5), 750-756 (1993-11-01)
Five commercially available devices for performing prick skin testing were compared for reproducibility, patient acceptance, occurrence of false-negative skin test results, and size distribution of reactions at the negative control sites. Reproducibility of skin testing with 10 mg/ml histamine base
L Shall et al.
Acta dermato-venereologica, 72(6), 451-453 (1992-11-01)
This study examined cutaneous mast cell behaviour in 14 patients with chronic urticaria but no dermographism and 11 healthy controls, by measuring cutaneous weal and flare reactions evoked in response to intradermal challenge injections of 0.1 ml isotonic saline, histamine
P Marshik et al.
Chest, 115(1), 194-199 (1999-01-30)
To determine optimal storage conditions for histamine diphosphate (HDP) solutions used for bronchoprovocation. HDP was dissolved in buffered saline solution to concentrations of 0.125 to 16 mg/mL and stored in 3-mL unit dose syringes at different temperatures for varying lengths
Y Larivée et al.
The Journal of otolaryngology, 30(6), 319-323 (2002-01-05)
The objectives of this study were to evaluate the reproducibility of the nasal response to histamine with the acoustic rhinometer and to compare the responses in normal and rhinitic subjects. Our study comprised 10 normal and 10 rhinitic subjects. Each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service